Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGS20
Entrez Gene ID
8601
Gene Name
regulator of G-protein signaling 20
Gene Aliases
RGSZ1, ZGAP1, g(z)GAP, gz-GAP
Chromosome Location
Chr.8: 53851808 - 53959304 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286673.1 NP_001273602.1 3 - 4 495 66
NM_001286675.1 NP_001273604.1 2 - 3 346 66
NM_003702.4 NP_003693.2 3 - 4 451 66
NM_170587.3 NP_733466.1 4 - 5 840 66
NR_104579.1 2 - 3 302 66
XM_011517622.2 XP_011515924.1 3 - 4 759 66
GenBank mRNA AF060877.2 - 3 - 4 318 66
AF074979.1 - 2 - 3 235 66
AF366054.1 - 3 - 4 403 66
AF366055.1 - 2 - 3 254 66
AF366057.1 - 3 - 4 268 66
AF493940.1 - 2 - 3 235 66
AK094503.1 - 2 - 3 302 66
AY046538.1 - 4 - 5 840 66
BC015614.2 - 3 - 4 288 66
BC063490.1 - 3 - 4 441 66
JQ308180.1 - 3 - 4 307 66

View Full Product Details