Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46922173-46922680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP35
Entrez Gene ID
2909
Gene Name
Rho GTPase activating protein 35
Gene Aliases
GRF-1, GRLF1, P190-A, P190A, p190ARhoGAP, p190RhoGAP
UniGene
Hs.509447
Gene Chromosome Location
Chr.19: 46861002-47005077 on Build GRCh38
Transcripts
NM_004491.4 XM_011526873.2 XM_017026714.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
998630 c.3571G>T p.E1191* Chr.19: 46922246-46922246 on Build GRCh38
4645359 c.3499C>T p.R1167W Chr.19: 46922174-46922174 on Build GRCh38
4939800 c.3587A>T p.Q1196L Chr.19: 46922262-46922262 on Build GRCh38
7249852 c.3531G>C p.V1177V Chr.19: 46922206-46922206 on Build GRCh38
3378788 c.3528C>T p.S1176S Chr.19: 46922203-46922203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780290702 Chr.19: 46922461-46922461 on Build GRCh38
rs372184901 Chr.19: 46922296-46922296 on Build GRCh38
rs947575939 Chr.19: 46922360-46922360 on Build GRCh38
rs199604498 Chr.19: 46922339-46922339 on Build GRCh38
rs145412639 Chr.19: 46922233-46922233 on Build GRCh38
rs888365105 Chr.19: 46922458-46922458 on Build GRCh38
rs558796421 Chr.19: 46922403-46922403 on Build GRCh38
rs754434697 Chr.19: 46922201-46922201 on Build GRCh38
rs747272619 Chr.19: 46922656-46922656 on Build GRCh38
rs895461618 Chr.19: 46922450-46922451 on Build GRCh38
rs762593594 Chr.19: 46922363-46922363 on Build GRCh38
rs566685888 Chr.19: 46922283-46922283 on Build GRCh38
rs1006787896 Chr.19: 46922574-46922574 on Build GRCh38
rs909440259 Chr.19: 46922365-46922365 on Build GRCh38
rs1002949973 Chr.19: 46922425-46922425 on Build GRCh38
rs374385673 Chr.19: 46922235-46922235 on Build GRCh38
rs929427082 Chr.19: 46922349-46922349 on Build GRCh38
rs943787062 Chr.19: 46922314-46922314 on Build GRCh38
rs746041884 Chr.19: 46922262-46922262 on Build GRCh38
rs538829439 Chr.19: 46922399-46922399 on Build GRCh38
rs916985183 Chr.19: 46922638-46922638 on Build GRCh38
rs879053344 Chr.19: 46922521-46922521 on Build GRCh38
rs185857289 Chr.19: 46922320-46922320 on Build GRCh38
rs1011339636 Chr.19: 46922483-46922483 on Build GRCh38
rs766115509 Chr.19: 46922231-46922231 on Build GRCh38
rs182325660 Chr.19: 46922447-46922447 on Build GRCh38
rs1044399012 Chr.19: 46922396-46922396 on Build GRCh38
rs772169199 Chr.19: 46922264-46922264 on Build GRCh38
rs191054812 Chr.19: 46922379-46922379 on Build GRCh38
rs891879745 Chr.19: 46922468-46922468 on Build GRCh38
rs985060376 Chr.19: 46922286-46922286 on Build GRCh38
rs767332838 Chr.19: 46922391-46922391 on Build GRCh38
rs968957609 Chr.19: 46922584-46922587 on Build GRCh38
rs373396486 Chr.19: 46922439-46922439 on Build GRCh38
rs16980767 Chr.19: 46922451-46922451 on Build GRCh38
rs988819975 Chr.19: 46922553-46922553 on Build GRCh38
rs965453771 Chr.19: 46922606-46922606 on Build GRCh38
rs758004253 Chr.19: 46922225-46922225 on Build GRCh38
rs191109761 Chr.19: 46922587-46922587 on Build GRCh38
rs996236003 Chr.19: 46922515-46922515 on Build GRCh38
rs764859054 Chr.19: 46922205-46922205 on Build GRCh38
rs186062421 Chr.19: 46922488-46922488 on Build GRCh38
rs775844443 Chr.19: 46922288-46922288 on Build GRCh38
rs1005373718 Chr.19: 46922196-46922196 on Build GRCh38
rs371111690 Chr.19: 46922204-46922204 on Build GRCh38
rs767031011 Chr.19: 46922362-46922363 on Build GRCh38
rs1016797606 Chr.19: 46922202-46922202 on Build GRCh38
rs767381909 Chr.19: 46922423-46922423 on Build GRCh38
rs972435057 Chr.19: 46922636-46922636 on Build GRCh38
rs954720757 Chr.19: 46922550-46922550 on Build GRCh38
rs777224632 Chr.19: 46922305-46922305 on Build GRCh38
rs371769271 Chr.19: 46922351-46922351 on Build GRCh38
rs369413520 Chr.19: 46922395-46922395 on Build GRCh38
rs765918008 Chr.19: 46922375-46922375 on Build GRCh38
rs1030357152 Chr.19: 46922528-46922528 on Build GRCh38
rs16980976 Chr.19: 46922460-46922461 on Build GRCh38
rs754809756 Chr.19: 46922234-46922234 on Build GRCh38
rs937109264 Chr.19: 46922610-46922610 on Build GRCh38
rs1020995590 Chr.19: 46922561-46922561 on Build GRCh38
rs1043227094 Chr.19: 46922369-46922369 on Build GRCh38
rs747302292 Chr.19: 46922293-46922293 on Build GRCh38
rs953949211 Chr.19: 46922511-46922511 on Build GRCh38
rs570213019 Chr.19: 46922386-46922386 on Build GRCh38
rs749571545 Chr.19: 46922367-46922368 on Build GRCh38
rs757285898 Chr.19: 46922244-46922244 on Build GRCh38
rs762418967 Chr.19: 46922313-46922313 on Build GRCh38
rs201142084 Chr.19: 46922243-46922243 on Build GRCh38
rs527979221 Chr.19: 46922216-46922216 on Build GRCh38
rs778967636 Chr.19: 46922248-46922248 on Build GRCh38

View Full Product Details