Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47673463-47673881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLTSCR1
Entrez Gene ID
29998
Gene Name
glioma tumor suppressor candidate region gene 1
Gene Aliases
-
UniGene
Hs.97244
Gene Chromosome Location
Chr.19: 47608196-47703277 on Build GRCh38
Transcripts
NM_015711.3 XM_005258833.4 XM_006723180.3 XM_011526883.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
293234 c.33C>T p.D11D Chr.19: 47673607-47673607 on Build GRCh38
4496198 c.46C>T p.P16S Chr.19: 47673724-47673724 on Build GRCh38
1129775 c.63C>A p.D21E Chr.19: 47673741-47673741 on Build GRCh38
3106578 c.34delG p.V12fs*1 Chr.19: 47673608-47673608 on Build GRCh38
5136604 c.41+9C>T p.? Chr.19: 47673624-47673624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201401494 Chr.19: 47673542-47673542 on Build GRCh38
rs370023070 Chr.19: 47673775-47673775 on Build GRCh38
rs763395550 Chr.19: 47673735-47673735 on Build GRCh38
rs749442299 Chr.19: 47673789-47673789 on Build GRCh38
rs779323268 Chr.19: 47673564-47673564 on Build GRCh38
rs777874220 Chr.19: 47673624-47673624 on Build GRCh38
rs987877285 Chr.19: 47673820-47673820 on Build GRCh38
rs554263925 Chr.19: 47673803-47673803 on Build GRCh38
rs538599147 Chr.19: 47673696-47673696 on Build GRCh38
rs889815178 Chr.19: 47673703-47673704 on Build GRCh38
rs372297380 Chr.19: 47673659-47673659 on Build GRCh38
rs780792935 Chr.19: 47673558-47673558 on Build GRCh38
rs371608441 Chr.19: 47673686-47673686 on Build GRCh38
rs746724443 Chr.19: 47673659-47673660 on Build GRCh38
rs773749290 Chr.19: 47673650-47673650 on Build GRCh38
rs759391445 Chr.19: 47673807-47673807 on Build GRCh38
rs866590723 Chr.19: 47673854-47673854 on Build GRCh38
rs772315357 Chr.19: 47673627-47673627 on Build GRCh38
rs773695952 Chr.19: 47673726-47673726 on Build GRCh38
rs754519998 Chr.19: 47673658-47673658 on Build GRCh38
rs745321979 Chr.19: 47673797-47673798 on Build GRCh38
rs769798555 Chr.19: 47673804-47673807 on Build GRCh38
rs755964864 Chr.19: 47673574-47673574 on Build GRCh38
rs768930906 Chr.19: 47673642-47673642 on Build GRCh38
rs377382503 Chr.19: 47673784-47673784 on Build GRCh38
rs374305375 Chr.19: 47673530-47673530 on Build GRCh38
rs199550150 Chr.19: 47673738-47673738 on Build GRCh38
rs770210656 Chr.19: 47673723-47673723 on Build GRCh38
rs943310006 Chr.19: 47673688-47673688 on Build GRCh38
rs775847054 Chr.19: 47673631-47673631 on Build GRCh38
rs116674006 Chr.19: 47673698-47673698 on Build GRCh38
rs759106800 Chr.19: 47673769-47673769 on Build GRCh38
rs776824884 Chr.19: 47673647-47673647 on Build GRCh38
rs913735359 Chr.19: 47673836-47673836 on Build GRCh38
rs375509450 Chr.19: 47673697-47673697 on Build GRCh38
rs745919921 Chr.19: 47673625-47673625 on Build GRCh38
rs1007232254 Chr.19: 47673498-47673498 on Build GRCh38
rs202093781 Chr.19: 47673720-47673720 on Build GRCh38
rs374601970 Chr.19: 47673780-47673780 on Build GRCh38
rs538465957 Chr.19: 47673808-47673808 on Build GRCh38
rs751281562 Chr.19: 47673547-47673547 on Build GRCh38
rs1019517824 Chr.19: 47673514-47673514 on Build GRCh38
rs939124186 Chr.19: 47673635-47673635 on Build GRCh38
rs1041401801 Chr.19: 47673737-47673737 on Build GRCh38
rs765931821 Chr.19: 47673654-47673654 on Build GRCh38
rs543412652 Chr.19: 47673842-47673842 on Build GRCh38
rs775327487 Chr.19: 47673808-47673809 on Build GRCh38
rs751094127 Chr.19: 47673657-47673657 on Build GRCh38
rs758535837 Chr.19: 47673673-47673673 on Build GRCh38
rs200303031 Chr.19: 47673671-47673671 on Build GRCh38
rs1016597804 Chr.19: 47673550-47673550 on Build GRCh38
rs755984704 Chr.19: 47673665-47673665 on Build GRCh38
rs563176294 Chr.19: 47673848-47673848 on Build GRCh38
rs368733953 Chr.19: 47673551-47673551 on Build GRCh38
rs762447003 Chr.19: 47673652-47673652 on Build GRCh38
rs770641055 Chr.19: 47673794-47673815 on Build GRCh38
rs747979687 Chr.19: 47673563-47673563 on Build GRCh38
rs776293156 Chr.19: 47673796-47673797 on Build GRCh38
rs150704505 Chr.19: 47673814-47673814 on Build GRCh38
rs960424294 Chr.19: 47673819-47673819 on Build GRCh38
rs200233936 Chr.19: 47673756-47673756 on Build GRCh38
rs536667799 Chr.19: 47673648-47673648 on Build GRCh38
rs377449854 Chr.19: 47673510-47673510 on Build GRCh38
rs753697249 Chr.19: 47673782-47673782 on Build GRCh38
rs969495215 Chr.19: 47673594-47673594 on Build GRCh38
rs375606918 Chr.19: 47673774-47673774 on Build GRCh38
rs952317181 Chr.19: 47673539-47673539 on Build GRCh38
rs754994318 Chr.19: 47673541-47673541 on Build GRCh38
rs367816725 Chr.19: 47673683-47673683 on Build GRCh38
rs747249197 Chr.19: 47673636-47673636 on Build GRCh38
rs373675820 Chr.19: 47673672-47673672 on Build GRCh38
rs755347130 Chr.19: 47673550-47673550 on Build GRCh38
rs778933932 Chr.19: 47673786-47673786 on Build GRCh38

View Full Product Details