The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320100.1 | NP_001307029.1 | 1 - 2 | 98 | 56 |
NM_001320101.1 | NP_001307030.1 | 1 - 2 | 98 | 56 | |
NM_003805.4 | NP_003796.1 | 1 - 2 | 98 | 56 | |
NR_135147.1 | 1 - 2 | 98 | 56 | ||
XM_017020142.1 | XP_016875631.1 | 1 - 2 | 127 | 56 | |
XM_017020143.1 | XP_016875632.1 | 1 - 2 | 121 | 56 | |
XM_017020144.1 | XP_016875633.1 | 1 - 2 | 121 | 56 | |
XM_017020145.1 | XP_016875634.1 | 1 - 2 | 120 | 56 | |
GenBank mRNA | AI791547.1 | - | 1 - 2 | 92 | 56 |
AK294986.1 | - | 1 - 2 | 86 | 56 | |
AK296225.1 | - | 1 - 2 | 96 | 56 | |
AK314861.1 | - | 1 - 2 | 78 | 56 | |
BC017042.1 | - | 1 - 2 | 70 | 56 | |
U79115.1 | - | 1 - 2 | 90 | 56 | |
U84388.1 | - | 1 - 2 | 27 | 56 |