Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50655968-50656473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf81
Entrez Gene ID
342918
Gene Name
chromosome 19 open reading frame 81
Gene Aliases
-
UniGene
Hs.531650
Gene Chromosome Location
Chr.19: 50649445-50659310 on Build GRCh38
Transcripts
NM_001195076.1 XM_011526910.2 XM_011527391.2

Gene Symbol
SYT3
Entrez Gene ID
84258
Gene Name
synaptotagmin 3
Gene Aliases
SytIII
UniGene
Hs.515554
Gene Chromosome Location
Chr.19: 50621977-50658127 on Build GRCh38
Transcripts
NM_001195076.1 XM_011526910.2 XM_011527391.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6003987 c.168A>G p.A56A Chr.19: 50656253-50656253 on Build GRCh38
5765542 c.197C>T p.P66L Chr.19: 50656282-50656282 on Build GRCh38
4591754 c.229T>G p.S77A Chr.19: 50656314-50656314 on Build GRCh38
5547374 c.237_242delCCTCTG p.L82_C83delLC Chr.19: 50656322-50656327 on Build GRCh38
3389243 c.146A>C p.Q49P Chr.19: 50656231-50656231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765432133 Chr.19: 50656375-50656376 on Build GRCh38
rs148726014 Chr.19: 50656335-50656335 on Build GRCh38
rs187877892 Chr.19: 50656111-50656111 on Build GRCh38
rs4801844 Chr.19: 50656314-50656314 on Build GRCh38
rs1021591390 Chr.19: 50656105-50656105 on Build GRCh38
rs751857046 Chr.19: 50656120-50656120 on Build GRCh38
rs908824661 Chr.19: 50656272-50656272 on Build GRCh38
rs558403135 Chr.19: 50656094-50656094 on Build GRCh38
rs934425304 Chr.19: 50656361-50656361 on Build GRCh38
rs749206263 Chr.19: 50656285-50656285 on Build GRCh38
rs530632779 Chr.19: 50656240-50656240 on Build GRCh38
rs192383921 Chr.19: 50656074-50656074 on Build GRCh38
rs915591512 Chr.19: 50655992-50655992 on Build GRCh38
rs117025012 Chr.19: 50656245-50656245 on Build GRCh38
rs1011492524 Chr.19: 50656099-50656099 on Build GRCh38
rs542605854 Chr.19: 50656062-50656062 on Build GRCh38
rs959581840 Chr.19: 50656260-50656260 on Build GRCh38
rs903010542 Chr.19: 50656032-50656032 on Build GRCh38
rs540911652 Chr.19: 50656098-50656098 on Build GRCh38
rs899174049 Chr.19: 50656166-50656166 on Build GRCh38
rs747308321 Chr.19: 50656428-50656428 on Build GRCh38
rs545205158 Chr.19: 50656191-50656191 on Build GRCh38
rs940324471 Chr.19: 50656273-50656273 on Build GRCh38
rs142260724 Chr.19: 50656364-50656364 on Build GRCh38
rs144243088 Chr.19: 50656290-50656290 on Build GRCh38
rs888949338 Chr.19: 50656373-50656373 on Build GRCh38
rs1007445473 Chr.19: 50656122-50656122 on Build GRCh38
rs978907970 Chr.19: 50656322-50656322 on Build GRCh38
rs571994021 Chr.19: 50656362-50656362 on Build GRCh38
rs984316344 Chr.19: 50656259-50656259 on Build GRCh38
rs569394144 Chr.19: 50656387-50656387 on Build GRCh38
rs756141833 Chr.19: 50656236-50656236 on Build GRCh38
rs977462844 Chr.19: 50656159-50656159 on Build GRCh38
rs991076028 Chr.19: 50656278-50656278 on Build GRCh38
rs1042813272 Chr.19: 50656025-50656025 on Build GRCh38
rs1051518787 Chr.19: 50656403-50656403 on Build GRCh38
rs1036511187 Chr.19: 50656064-50656064 on Build GRCh38
rs867757230 Chr.19: 50656298-50656298 on Build GRCh38
rs887436354 Chr.19: 50656052-50656052 on Build GRCh38
rs991582347 Chr.19: 50656213-50656213 on Build GRCh38
rs868829154 Chr.19: 50656271-50656271 on Build GRCh38
rs765677846 Chr.19: 50656070-50656070 on Build GRCh38
rs755142683 Chr.19: 50656152-50656152 on Build GRCh38
rs57195892 Chr.19: 50656212-50656212 on Build GRCh38
rs745596786 Chr.19: 50656352-50656352 on Build GRCh38
rs538813754 Chr.19: 50656363-50656363 on Build GRCh38
rs192093645 Chr.19: 50656158-50656158 on Build GRCh38
rs185628472 Chr.19: 50656095-50656095 on Build GRCh38
rs933065966 Chr.19: 50656320-50656320 on Build GRCh38
rs762384798 Chr.19: 50656035-50656035 on Build GRCh38
rs575701172 Chr.19: 50656013-50656013 on Build GRCh38
rs924409330 Chr.19: 50656328-50656328 on Build GRCh38
rs1050206561 Chr.19: 50656353-50656353 on Build GRCh38
rs1011439775 Chr.19: 50656390-50656390 on Build GRCh38
rs778504965 Chr.19: 50656116-50656116 on Build GRCh38

View Full Product Details