Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CPT1B
- Entrez Gene ID
-
1375
- Gene Name
- carnitine palmitoyltransferase 1B
- Gene Aliases
-
CPT1-M, CPT1M, CPTI, CPTI-M, M-CPT1, MCCPT1, MCPT1
- Chromosome Location
-
Chr.22: 50568861 - 50578667 on Build GRCh38
- Gene Symbol
- CHKB-CPT1B
- Entrez Gene ID
-
386593
- Gene Name
- CHKB-CPT1B readthrough (NMD candidate)
- Gene Aliases
-
CHKL-CPT1B
- Chromosome Location
-
Chr.22: 50568861 - 50582999 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027928.2 |
|
17 - 18 |
2353 |
67 |
GenBank mRNA |
AB051457.1 |
- |
14 - 15 |
2991 |
67 |
BC131570.1 |
- |
9 - 10 |
1134 |
67 |
View Full Product Details