Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9587980-9588492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YWHAQ
Entrez Gene ID
10971
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein theta
Gene Aliases
14-3-3, 1C5, HS1
UniGene
Hs.74405
Gene Chromosome Location
Chr.2: 9583967-9631055 on Build GRCh38
Transcripts
NM_006826.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4583711 c.474A>G p.K158K Chr.2: 9588273-9588273 on Build GRCh38
4096408 c.499C>T p.R167C Chr.2: 9588248-9588248 on Build GRCh38
4096407 c.564C>T p.A188A Chr.2: 9588183-9588183 on Build GRCh38
5617145 c.582+8G>T p.? Chr.2: 9588157-9588157 on Build GRCh38
3799282 c.574G>A p.A192T Chr.2: 9588173-9588173 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755703871 Chr.2: 9588240-9588240 on Build GRCh38
rs746259702 Chr.2: 9588142-9588142 on Build GRCh38
rs537900235 Chr.2: 9588075-9588075 on Build GRCh38
rs917812194 Chr.2: 9588155-9588155 on Build GRCh38
rs762976369 Chr.2: 9588177-9588177 on Build GRCh38
rs375412137 Chr.2: 9588349-9588349 on Build GRCh38
rs369032698 Chr.2: 9588135-9588135 on Build GRCh38
rs367832498 Chr.2: 9588247-9588247 on Build GRCh38
rs903147610 Chr.2: 9588432-9588433 on Build GRCh38
rs774221614 Chr.2: 9588308-9588308 on Build GRCh38
rs751249633 Chr.2: 9588318-9588318 on Build GRCh38
rs531332618 Chr.2: 9588433-9588433 on Build GRCh38
rs555932570 Chr.2: 9588042-9588042 on Build GRCh38
rs750901996 Chr.2: 9588361-9588361 on Build GRCh38
rs371354151 Chr.2: 9588293-9588293 on Build GRCh38
rs772387887 Chr.2: 9588263-9588263 on Build GRCh38
rs952011580 Chr.2: 9588395-9588395 on Build GRCh38
rs531964742 Chr.2: 9588049-9588052 on Build GRCh38
rs557983507 Chr.2: 9588102-9588102 on Build GRCh38
rs780770187 Chr.2: 9588352-9588352 on Build GRCh38
rs767340019 Chr.2: 9588195-9588195 on Build GRCh38
rs998233802 Chr.2: 9588121-9588121 on Build GRCh38
rs929883882 Chr.2: 9588024-9588024 on Build GRCh38
rs749806408 Chr.2: 9588122-9588122 on Build GRCh38
rs984896851 Chr.2: 9588415-9588415 on Build GRCh38
rs947719761 Chr.2: 9588164-9588164 on Build GRCh38
rs746377347 Chr.2: 9588258-9588258 on Build GRCh38
rs199515928 Chr.2: 9588275-9588275 on Build GRCh38
rs780502121 Chr.2: 9588264-9588264 on Build GRCh38
rs537211068 Chr.2: 9588059-9588059 on Build GRCh38
rs754783750 Chr.2: 9588192-9588192 on Build GRCh38
rs984864200 Chr.2: 9588090-9588090 on Build GRCh38
rs201265459 Chr.2: 9588339-9588339 on Build GRCh38
rs200884745 Chr.2: 9588251-9588251 on Build GRCh38
rs927603470 Chr.2: 9588270-9588270 on Build GRCh38
rs751866146 Chr.2: 9588389-9588389 on Build GRCh38
rs752439118 Chr.2: 9588332-9588332 on Build GRCh38
rs777288889 Chr.2: 9588245-9588245 on Build GRCh38
rs761844291 Chr.2: 9588297-9588297 on Build GRCh38
rs751488523 Chr.2: 9588189-9588189 on Build GRCh38
rs564769446 Chr.2: 9588446-9588446 on Build GRCh38
rs1029031353 Chr.2: 9588378-9588378 on Build GRCh38
rs775916718 Chr.2: 9588149-9588149 on Build GRCh38
rs1023423895 Chr.2: 9588243-9588243 on Build GRCh38
rs551157053 Chr.2: 9588068-9588068 on Build GRCh38
rs769900540 Chr.2: 9588305-9588305 on Build GRCh38
rs373146755 Chr.2: 9588137-9588137 on Build GRCh38
rs776796788 Chr.2: 9588169-9588169 on Build GRCh38
rs941235411 Chr.2: 9588387-9588387 on Build GRCh38
rs749924341 Chr.2: 9588127-9588128 on Build GRCh38
rs144863616 Chr.2: 9588273-9588273 on Build GRCh38
rs201487177 Chr.2: 9588276-9588276 on Build GRCh38
rs948035520 Chr.2: 9588447-9588447 on Build GRCh38
rs934055641 Chr.2: 9588411-9588411 on Build GRCh38
rs1000606168 Chr.2: 9588248-9588248 on Build GRCh38
rs759450776 Chr.2: 9588323-9588323 on Build GRCh38
rs752349662 Chr.2: 9588222-9588222 on Build GRCh38
rs1010382306 Chr.2: 9588336-9588336 on Build GRCh38
rs1052123960 Chr.2: 9588307-9588307 on Build GRCh38
rs907935681 Chr.2: 9588431-9588431 on Build GRCh38
rs150523147 Chr.2: 9588178-9588178 on Build GRCh38
rs768956586 Chr.2: 9588283-9588283 on Build GRCh38
rs72777035 Chr.2: 9588448-9588448 on Build GRCh38
rs999086390 Chr.2: 9588145-9588145 on Build GRCh38
rs67724256 Chr.2: 9588445-9588445 on Build GRCh38
rs187591182 Chr.2: 9588148-9588148 on Build GRCh38
rs76278872 Chr.2: 9588340-9588340 on Build GRCh38
rs899904494 Chr.2: 9588120-9588120 on Build GRCh38
rs201336910 Chr.2: 9588150-9588150 on Build GRCh38
rs767312364 Chr.2: 9588325-9588325 on Build GRCh38
rs940486137 Chr.2: 9588141-9588141 on Build GRCh38
rs752731976 Chr.2: 9588058-9588058 on Build GRCh38
rs758884857 Chr.2: 9588377-9588377 on Build GRCh38

View Full Product Details