Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9630080-9630583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YWHAQ
Entrez Gene ID
10971
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein theta
Gene Aliases
14-3-3, 1C5, HS1
UniGene
Hs.74405
Gene Chromosome Location
Chr.2: 9583967-9631055 on Build GRCh38
Transcripts
NM_006826.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1024027 c.252G>T p.E84D Chr.2: 9630201-9630201 on Build GRCh38
6824612 c.191G>T p.S64I Chr.2: 9630262-9630262 on Build GRCh38
4464962 c.135C>T p.S45S Chr.2: 9630318-9630318 on Build GRCh38
6824608 c.206C>A p.T69N Chr.2: 9630247-9630247 on Build GRCh38
6584199 c.270G>A p.L90L Chr.2: 9630183-9630183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751552379 Chr.2: 9630495-9630495 on Build GRCh38
rs755954786 Chr.2: 9630472-9630472 on Build GRCh38
rs960357310 Chr.2: 9630272-9630272 on Build GRCh38
rs979488247 Chr.2: 9630467-9630467 on Build GRCh38
rs763241227 Chr.2: 9630541-9630541 on Build GRCh38
rs372377118 Chr.2: 9630130-9630130 on Build GRCh38
rs745504240 Chr.2: 9630479-9630479 on Build GRCh38
rs749499495 Chr.2: 9630242-9630242 on Build GRCh38
rs746966183 Chr.2: 9630263-9630263 on Build GRCh38
rs1015795021 Chr.2: 9630536-9630536 on Build GRCh38
rs559702166 Chr.2: 9630475-9630475 on Build GRCh38
rs756561669 Chr.2: 9630340-9630340 on Build GRCh38
rs375411374 Chr.2: 9630483-9630483 on Build GRCh38
rs774663201 Chr.2: 9630477-9630478 on Build GRCh38
rs1027369792 Chr.2: 9630210-9630210 on Build GRCh38
rs960447911 Chr.2: 9630542-9630542 on Build GRCh38
rs755484324 Chr.2: 9630213-9630213 on Build GRCh38
rs931465361 Chr.2: 9630523-9630523 on Build GRCh38
rs11544465 Chr.2: 9630236-9630236 on Build GRCh38
rs776483508 Chr.2: 9630481-9630481 on Build GRCh38
rs550380937 Chr.2: 9630557-9630557 on Build GRCh38
rs765579544 Chr.2: 9630487-9630487 on Build GRCh38
rs764174434 Chr.2: 9630501-9630501 on Build GRCh38
rs777649657 Chr.2: 9630381-9630381 on Build GRCh38
rs748029699 Chr.2: 9630369-9630369 on Build GRCh38
rs773424346 Chr.2: 9630489-9630489 on Build GRCh38
rs915919557 Chr.2: 9630399-9630399 on Build GRCh38
rs769631927 Chr.2: 9630373-9630373 on Build GRCh38
rs966744955 Chr.2: 9630106-9630106 on Build GRCh38
rs762606877 Chr.2: 9630177-9630177 on Build GRCh38
rs766499891 Chr.2: 9630492-9630492 on Build GRCh38
rs748020959 Chr.2: 9630312-9630312 on Build GRCh38
rs1130874 Chr.2: 9630484-9630484 on Build GRCh38
rs750464603 Chr.2: 9630356-9630356 on Build GRCh38
rs765485923 Chr.2: 9630459-9630459 on Build GRCh38
rs763024118 Chr.2: 9630491-9630491 on Build GRCh38
rs545839277 Chr.2: 9630462-9630462 on Build GRCh38
rs777741592 Chr.2: 9630474-9630474 on Build GRCh38
rs368778993 Chr.2: 9630108-9630108 on Build GRCh38
rs778901859 Chr.2: 9630253-9630253 on Build GRCh38
rs1039072577 Chr.2: 9630544-9630544 on Build GRCh38
rs1020766289 Chr.2: 9630382-9630382 on Build GRCh38
rs768534041 Chr.2: 9630267-9630267 on Build GRCh38
rs774805872 Chr.2: 9630481-9630481 on Build GRCh38
rs377250330 Chr.2: 9630342-9630342 on Build GRCh38
rs778991719 Chr.2: 9630355-9630355 on Build GRCh38
rs1017557119 Chr.2: 9630354-9630354 on Build GRCh38
rs1027421128 Chr.2: 9630504-9630504 on Build GRCh38
rs886220286 Chr.2: 9630510-9630510 on Build GRCh38
rs372978538 Chr.2: 9630336-9630336 on Build GRCh38
rs777744668 Chr.2: 9630127-9630127 on Build GRCh38
rs926010256 Chr.2: 9630471-9630471 on Build GRCh38
rs746945960 Chr.2: 9630140-9630140 on Build GRCh38
rs761020810 Chr.2: 9630438-9630438 on Build GRCh38
rs776325430 Chr.2: 9630281-9630281 on Build GRCh38
rs762624973 Chr.2: 9630318-9630318 on Build GRCh38
rs773471715 Chr.2: 9630461-9630466 on Build GRCh38
rs11544464 Chr.2: 9630463-9630463 on Build GRCh38
rs141887759 Chr.2: 9630368-9630368 on Build GRCh38
rs763733202 Chr.2: 9630479-9630480 on Build GRCh38
rs756308327 Chr.2: 9630119-9630119 on Build GRCh38
rs760535815 Chr.2: 9630478-9630479 on Build GRCh38
rs369469100 Chr.2: 9630552-9630552 on Build GRCh38
rs1037601336 Chr.2: 9630178-9630178 on Build GRCh38
rs764536655 Chr.2: 9630454-9630454 on Build GRCh38
rs982921555 Chr.2: 9630257-9630257 on Build GRCh38
rs1048828426 Chr.2: 9630532-9630532 on Build GRCh38
rs769460753 Chr.2: 9630315-9630315 on Build GRCh38
rs201957113 Chr.2: 9630330-9630330 on Build GRCh38
rs755053877 Chr.2: 9630497-9630497 on Build GRCh38
rs202138500 Chr.2: 9630234-9630234 on Build GRCh38
rs200302461 Chr.2: 9630460-9630461 on Build GRCh38
rs757416206 Chr.2: 9630353-9630353 on Build GRCh38
rs773949858 Chr.2: 9630326-9630326 on Build GRCh38
rs778766840 Chr.2: 9630477-9630477 on Build GRCh38
rs748332272 Chr.2: 9630116-9630116 on Build GRCh38
rs758568866 Chr.2: 9630465-9630465 on Build GRCh38
rs918734637 Chr.2: 9630517-9630517 on Build GRCh38
rs752112912 Chr.2: 9630208-9630208 on Build GRCh38
rs754157288 Chr.2: 9630456-9630456 on Build GRCh38
rs776433787 Chr.2: 9630165-9630165 on Build GRCh38
rs901767378 Chr.2: 9630545-9630545 on Build GRCh38
rs369852702 Chr.2: 9630176-9630176 on Build GRCh38
rs781724742 Chr.2: 9630110-9630122 on Build GRCh38
rs756400871 Chr.2: 9630232-9630232 on Build GRCh38
rs747591783 Chr.2: 9630550-9630550 on Build GRCh38
rs748970639 Chr.2: 9630390-9630390 on Build GRCh38
rs11544463 Chr.2: 9630520-9630520 on Build GRCh38
rs764907087 Chr.2: 9630174-9630174 on Build GRCh38
rs371355879 Chr.2: 9630152-9630152 on Build GRCh38
rs755457718 Chr.2: 9630109-9630109 on Build GRCh38
rs752690192 Chr.2: 9630470-9630470 on Build GRCh38
rs1039329287 Chr.2: 9630490-9630490 on Build GRCh38
rs770978736 Chr.2: 9630136-9630136 on Build GRCh38
rs774239040 Chr.2: 9630199-9630199 on Build GRCh38
rs770667690 Chr.2: 9630393-9630393 on Build GRCh38
rs754065297 Chr.2: 9630351-9630351 on Build GRCh38
rs199958721 Chr.2: 9630214-9630214 on Build GRCh38
rs375839408 Chr.2: 9630133-9630133 on Build GRCh38
rs753064140 Chr.2: 9630225-9630225 on Build GRCh38
rs997814161 Chr.2: 9630552-9630556 on Build GRCh38
rs539717165 Chr.2: 9630300-9630300 on Build GRCh38
rs369055092 Chr.2: 9630144-9630144 on Build GRCh38
rs372545955 Chr.2: 9630182-9630182 on Build GRCh38
rs776134213 Chr.2: 9630424-9630424 on Build GRCh38
rs757395602 Chr.2: 9630246-9630246 on Build GRCh38
rs748667930 Chr.2: 9630110-9630110 on Build GRCh38
rs762217816 Chr.2: 9630485-9630485 on Build GRCh38
rs758380692 Chr.2: 9630362-9630362 on Build GRCh38
rs561528893 Chr.2: 9630516-9630516 on Build GRCh38
rs771615816 Chr.2: 9630480-9630480 on Build GRCh38
rs761396781 Chr.2: 9630169-9630169 on Build GRCh38
rs948687072 Chr.2: 9630437-9630437 on Build GRCh38
rs368512939 Chr.2: 9630137-9630137 on Build GRCh38
rs779927922 Chr.2: 9630468-9630468 on Build GRCh38
rs760149820 Chr.2: 9630328-9630328 on Build GRCh38
rs781552947 Chr.2: 9630113-9630113 on Build GRCh38
rs368668324 Chr.2: 9630414-9630414 on Build GRCh38
rs774114759 Chr.2: 9630395-9630395 on Build GRCh38
rs541763260 Chr.2: 9630512-9630512 on Build GRCh38

View Full Product Details