The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277161.1 | NP_001264090.1 | 4 - 5 | 1487 | 95 |
NM_004745.4 | NP_004736.2 | 4 - 5 | 1487 | 95 | |
GenBank mRNA | AF009204.1 | - | 3 - 4 | 1300 | 95 |
AF119817.1 | - | 4 - 5 | 1472 | 95 | |
AF119818.1 | - | 4 - 5 | 1487 | 95 | |
BC141862.1 | - | 3 - 4 | 1403 | 95 | |
KJ534823.1 | - | 3 - 4 | 1353 | 95 |