The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105250.2 | NP_001098720.1 | 6 - 7 | 2030 | 90 |
NM_004796.5 | NP_004787.2 | 16 - 17 | 3393 | 90 | |
XM_011537372.1 | XP_011535674.1 | 22 - 23 | 5133 | 90 | |
XM_011537373.1 | XP_011535675.1 | 21 - 22 | 5043 | 90 | |
XM_017021803.1 | XP_016877292.1 | 21 - 22 | 5112 | 90 | |
XM_017021804.1 | XP_016877293.1 | 21 - 22 | 5106 | 90 | |
XM_017021805.1 | XP_016877294.1 | 20 - 21 | 5097 | 90 | |
GenBank mRNA | AB018286.1 | - | 16 - 17 | 3374 | 90 |
BC142649.1 | - | 6 - 7 | 1463 | 90 | |
BC150194.1 | - | 6 - 7 | 1463 | 90 | |
BC152457.1 | - | 16 - 17 | 3374 | 90 |