The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004869.3 | NP_004860.2 | 3 - 4 | 556 | 58 |
XM_011526268.2 | XP_011524570.1 | 3 - 4 | 556 | 58 | |
GenBank mRNA | AF038960.1 | - | 3 - 4 | 485 | 58 |
AF195514.1 | - | 3 - 4 | 497 | 58 | |
AK290932.1 | - | 3 - 4 | 500 | 58 | |
AK291253.1 | - | 3 - 4 | 454 | 58 | |
AK308892.1 | - | 3 - 4 | 520 | 58 | |
AY232629.1 | - | 3 - 4 | 306 | 58 | |
BC039574.1 | - | 3 - 4 | 463 | 58 | |
DA306069.1 | - | 3 - 4 | 557 | 58 |