Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27123699-27124195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD1
Entrez Gene ID
84696
Gene Name
abhydrolase domain containing 1
Gene Aliases
LABH1
UniGene
Hs.375791
Gene Chromosome Location
Chr.2: 27123780-27130812 on Build GRCh38
Transcripts
NM_032604.3 XM_011533135.2 XM_011533136.2 XM_011533137.2 XM_011533138.2 XM_017005113.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6256552 c.70G>T p.A24S Chr.2: 27124018-27124018 on Build GRCh38
4093467 c.8G>A p.S3N Chr.2: 27123956-27123956 on Build GRCh38
7106883 c.87G>C p.L29F Chr.2: 27124035-27124035 on Build GRCh38
5571078 c.73G>A p.V25I Chr.2: 27124021-27124021 on Build GRCh38
4583428 c.107T>C p.V36A Chr.2: 27124055-27124055 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758304621 Chr.2: 27124052-27124052 on Build GRCh38
rs778515207 Chr.2: 27124046-27124046 on Build GRCh38
rs563458977 Chr.2: 27123752-27123752 on Build GRCh38
rs199784100 Chr.2: 27123976-27123976 on Build GRCh38
rs946306555 Chr.2: 27123720-27123720 on Build GRCh38
rs756838881 Chr.2: 27124045-27124045 on Build GRCh38
rs753563733 Chr.2: 27124087-27124087 on Build GRCh38
rs1026872568 Chr.2: 27123825-27123825 on Build GRCh38
rs748571832 Chr.2: 27124105-27124105 on Build GRCh38
rs776060710 Chr.2: 27124080-27124080 on Build GRCh38
rs368108479 Chr.2: 27123942-27123942 on Build GRCh38
rs992111282 Chr.2: 27124138-27124138 on Build GRCh38
rs761211351 Chr.2: 27124134-27124134 on Build GRCh38
rs372557192 Chr.2: 27124097-27124097 on Build GRCh38
rs962742300 Chr.2: 27123849-27123849 on Build GRCh38
rs777545125 Chr.2: 27123966-27123966 on Build GRCh38
rs1042613966 Chr.2: 27123754-27123754 on Build GRCh38
rs775873241 Chr.2: 27124133-27124133 on Build GRCh38
rs764581353 Chr.2: 27124140-27124140 on Build GRCh38
rs768037800 Chr.2: 27124021-27124021 on Build GRCh38
rs777171342 Chr.2: 27124155-27124155 on Build GRCh38
rs965109842 Chr.2: 27123843-27123843 on Build GRCh38
rs1029907233 Chr.2: 27123827-27123827 on Build GRCh38
rs756927179 Chr.2: 27124089-27124089 on Build GRCh38
rs978490867 Chr.2: 27124092-27124092 on Build GRCh38
rs895941613 Chr.2: 27123791-27123791 on Build GRCh38
rs879874333 Chr.2: 27123760-27123760 on Build GRCh38
rs1043292467 Chr.2: 27123726-27123726 on Build GRCh38
rs763012248 Chr.2: 27123928-27123928 on Build GRCh38
rs777633358 Chr.2: 27124042-27124042 on Build GRCh38
rs994101217 Chr.2: 27123821-27123821 on Build GRCh38
rs749808560 Chr.2: 27123738-27123738 on Build GRCh38
rs775104040 Chr.2: 27123986-27123986 on Build GRCh38
rs1056350505 Chr.2: 27123755-27123755 on Build GRCh38
rs747908091 Chr.2: 27124116-27124116 on Build GRCh38
rs771193388 Chr.2: 27124123-27124123 on Build GRCh38
rs936511906 Chr.2: 27123989-27123990 on Build GRCh38
rs759639089 Chr.2: 27124076-27124076 on Build GRCh38
rs145553516 Chr.2: 27124044-27124044 on Build GRCh38
rs1036371222 Chr.2: 27123813-27123813 on Build GRCh38
rs756487979 Chr.2: 27124025-27124025 on Build GRCh38
rs997221422 Chr.2: 27123823-27123823 on Build GRCh38
rs771920338 Chr.2: 27123983-27123983 on Build GRCh38
rs878864406 Chr.2: 27123871-27123871 on Build GRCh38
rs978821322 Chr.2: 27123881-27123881 on Build GRCh38
rs751999519 Chr.2: 27123939-27123939 on Build GRCh38
rs575143127 Chr.2: 27124103-27124103 on Build GRCh38
rs746002110 Chr.2: 27124127-27124127 on Build GRCh38
rs768375869 Chr.2: 27124061-27124061 on Build GRCh38
rs966988102 Chr.2: 27123875-27123875 on Build GRCh38
rs950115579 Chr.2: 27123959-27123959 on Build GRCh38
rs976434689 Chr.2: 27123865-27123865 on Build GRCh38
rs753051527 Chr.2: 27124022-27124022 on Build GRCh38
rs1006747248 Chr.2: 27123840-27123840 on Build GRCh38
rs754791446 Chr.2: 27124106-27124106 on Build GRCh38
rs769312213 Chr.2: 27123909-27123912 on Build GRCh38
rs1018518655 Chr.2: 27123841-27123841 on Build GRCh38
rs752426472 Chr.2: 27123956-27123956 on Build GRCh38
rs764947361 Chr.2: 27124090-27124090 on Build GRCh38
rs142253735 Chr.2: 27123867-27123868 on Build GRCh38
rs746910638 Chr.2: 27124105-27124105 on Build GRCh38
rs541783608 Chr.2: 27123736-27123736 on Build GRCh38
rs763028743 Chr.2: 27124001-27124001 on Build GRCh38
rs767966501 Chr.2: 27124079-27124079 on Build GRCh38
rs765386098 Chr.2: 27123917-27123917 on Build GRCh38
rs202173691 Chr.2: 27123957-27123957 on Build GRCh38
rs879114072 Chr.2: 27123882-27123882 on Build GRCh38
rs181018386 Chr.2: 27123817-27123817 on Build GRCh38
rs556357900 Chr.2: 27123729-27123730 on Build GRCh38
rs146509505 Chr.2: 27124060-27124060 on Build GRCh38
rs531007063 Chr.2: 27123769-27123769 on Build GRCh38
rs369687036 Chr.2: 27124066-27124066 on Build GRCh38
rs764493559 Chr.2: 27124083-27124083 on Build GRCh38
rs762243198 Chr.2: 27124163-27124163 on Build GRCh38
rs376400996 Chr.2: 27123941-27123941 on Build GRCh38
rs766940652 Chr.2: 27123937-27123937 on Build GRCh38
rs766390056 Chr.2: 27124005-27124005 on Build GRCh38
rs769984653 Chr.2: 27124137-27124138 on Build GRCh38
rs752085651 Chr.2: 27124006-27124006 on Build GRCh38
rs749464726 Chr.2: 27124067-27124067 on Build GRCh38
rs945193785 Chr.2: 27123753-27123753 on Build GRCh38
rs780031793 Chr.2: 27124055-27124055 on Build GRCh38
rs771694746 Chr.2: 27123898-27123898 on Build GRCh38
rs546109785 Chr.2: 27123809-27123809 on Build GRCh38
rs1049512757 Chr.2: 27124008-27124008 on Build GRCh38
rs773421825 Chr.2: 27123997-27123997 on Build GRCh38
rs779862769 Chr.2: 27124104-27124104 on Build GRCh38
rs200272895 Chr.2: 27123904-27123904 on Build GRCh38
rs769973107 Chr.2: 27123995-27123995 on Build GRCh38
rs371837081 Chr.2: 27123918-27123918 on Build GRCh38
rs879179309 Chr.2: 27123878-27123878 on Build GRCh38
rs369113573 Chr.2: 27123733-27123734 on Build GRCh38
rs764868129 Chr.2: 27124164-27124164 on Build GRCh38
rs958993695 Chr.2: 27124108-27124108 on Build GRCh38
rs539496364 Chr.2: 27124124-27124124 on Build GRCh38
rs190780666 Chr.2: 27123785-27123785 on Build GRCh38
rs550557997 Chr.2: 27124051-27124051 on Build GRCh38
rs772120708 Chr.2: 27124130-27124130 on Build GRCh38
rs759921353 Chr.2: 27124019-27124019 on Build GRCh38
rs912920992 Chr.2: 27123867-27123867 on Build GRCh38
rs974117019 Chr.2: 27123850-27123850 on Build GRCh38
rs774334737 Chr.2: 27124074-27124074 on Build GRCh38
rs745731118 Chr.2: 27123980-27123980 on Build GRCh38
rs376010551 Chr.2: 27123892-27123892 on Build GRCh38
rs926735377 Chr.2: 27123883-27123883 on Build GRCh38
rs777245287 Chr.2: 27124040-27124041 on Build GRCh38
rs771032113 Chr.2: 27124073-27124073 on Build GRCh38
rs115289202 Chr.2: 27123831-27123831 on Build GRCh38
rs780926745 Chr.2: 27124109-27124109 on Build GRCh38
rs749947165 Chr.2: 27124091-27124091 on Build GRCh38
rs1056367190 Chr.2: 27123728-27123728 on Build GRCh38
rs926127462 Chr.2: 27123735-27123735 on Build GRCh38
rs372003324 Chr.2: 27123969-27123969 on Build GRCh38
rs375202801 Chr.2: 27123973-27123973 on Build GRCh38
rs888407789 Chr.2: 27124013-27124013 on Build GRCh38
rs760484404 Chr.2: 27123989-27123989 on Build GRCh38
rs745637487 Chr.2: 27123885-27123885 on Build GRCh38

View Full Product Details