Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27129675-27130174 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD1
Entrez Gene ID
84696
Gene Name
abhydrolase domain containing 1
Gene Aliases
LABH1
UniGene
Hs.375791
Gene Chromosome Location
Chr.2: 27123780-27130812 on Build GRCh38
Transcripts
NM_032604.3 XM_011533135.2 XM_011533136.2 XM_011533137.2 XM_011533138.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5973742 c.681_684delTCTG p.S229fs*39 Chr.2: 27129817-27129820 on Build GRCh38
7090747 c.645G>T p.Q215H Chr.2: 27129781-27129781 on Build GRCh38
374333 c.721C>A p.L241M Chr.2: 27129857-27129857 on Build GRCh38
6157667 c.682C>A p.L228M Chr.2: 27129818-27129818 on Build GRCh38
3743751 c.806T>C p.I269T Chr.2: 27130119-27130119 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760489485 Chr.2: 27129933-27129933 on Build GRCh38
rs770543449 Chr.2: 27129800-27129800 on Build GRCh38
rs139038618 Chr.2: 27129859-27129859 on Build GRCh38
rs373511955 Chr.2: 27129978-27129978 on Build GRCh38
rs867541474 Chr.2: 27130105-27130105 on Build GRCh38
rs768532603 Chr.2: 27130073-27130073 on Build GRCh38
rs772677093 Chr.2: 27129720-27129720 on Build GRCh38
rs926073398 Chr.2: 27130035-27130035 on Build GRCh38
rs781406854 Chr.2: 27129766-27129767 on Build GRCh38
rs192877845 Chr.2: 27129746-27129746 on Build GRCh38
rs759030905 Chr.2: 27129811-27129811 on Build GRCh38
rs71441057 Chr.2: 27130021-27130021 on Build GRCh38
rs759721270 Chr.2: 27130088-27130088 on Build GRCh38
rs887029388 Chr.2: 27129876-27129876 on Build GRCh38
rs146000122 Chr.2: 27129766-27129766 on Build GRCh38
rs761697867 Chr.2: 27129942-27129942 on Build GRCh38
rs138068203 Chr.2: 27130087-27130087 on Build GRCh38
rs770143523 Chr.2: 27129818-27129819 on Build GRCh38
rs772944608 Chr.2: 27129777-27129777 on Build GRCh38
rs751774268 Chr.2: 27129949-27129949 on Build GRCh38
rs765545178 Chr.2: 27130119-27130119 on Build GRCh38
rs200164733 Chr.2: 27129805-27129805 on Build GRCh38
rs762467417 Chr.2: 27129721-27129721 on Build GRCh38
rs768605892 Chr.2: 27129935-27129935 on Build GRCh38
rs922729542 Chr.2: 27129987-27129987 on Build GRCh38
rs1020995804 Chr.2: 27130010-27130010 on Build GRCh38
rs967649434 Chr.2: 27130028-27130028 on Build GRCh38
rs190083760 Chr.2: 27130052-27130052 on Build GRCh38
rs776645901 Chr.2: 27130075-27130075 on Build GRCh38
rs776615298 Chr.2: 27129936-27129936 on Build GRCh38
rs754481629 Chr.2: 27130139-27130139 on Build GRCh38
rs766714991 Chr.2: 27129947-27129947 on Build GRCh38
rs773309161 Chr.2: 27129874-27129895 on Build GRCh38
rs546593109 Chr.2: 27130076-27130076 on Build GRCh38
rs941292461 Chr.2: 27130050-27130050 on Build GRCh38
rs1033579262 Chr.2: 27129966-27129966 on Build GRCh38
rs775220730 Chr.2: 27129822-27129822 on Build GRCh38
rs757637123 Chr.2: 27130116-27130116 on Build GRCh38
rs778546503 Chr.2: 27129964-27129964 on Build GRCh38
rs778042196 Chr.2: 27129888-27129888 on Build GRCh38
rs778530056 Chr.2: 27129925-27129925 on Build GRCh38
rs753096530 Chr.2: 27129870-27129870 on Build GRCh38
rs763747273 Chr.2: 27129827-27129827 on Build GRCh38
rs926032954 Chr.2: 27129736-27129736 on Build GRCh38
rs185150956 Chr.2: 27129927-27129927 on Build GRCh38
rs748059020 Chr.2: 27130142-27130142 on Build GRCh38
rs370160762 Chr.2: 27129963-27129963 on Build GRCh38
rs779469800 Chr.2: 27130059-27130059 on Build GRCh38
rs761365496 Chr.2: 27130132-27130132 on Build GRCh38
rs747053287 Chr.2: 27129761-27129761 on Build GRCh38
rs908437480 Chr.2: 27130046-27130046 on Build GRCh38
rs765827051 Chr.2: 27129725-27129725 on Build GRCh38
rs201450092 Chr.2: 27129959-27129959 on Build GRCh38
rs141714250 Chr.2: 27129926-27129926 on Build GRCh38
rs926062082 Chr.2: 27129844-27129844 on Build GRCh38
rs760527669 Chr.2: 27129824-27129824 on Build GRCh38
rs748551971 Chr.2: 27129770-27129770 on Build GRCh38
rs751757700 Chr.2: 27129851-27129851 on Build GRCh38
rs746504266 Chr.2: 27130060-27130060 on Build GRCh38
rs144967995 Chr.2: 27129878-27129878 on Build GRCh38
rs376409416 Chr.2: 27130057-27130057 on Build GRCh38
rs955336505 Chr.2: 27130014-27130014 on Build GRCh38
rs758034669 Chr.2: 27130129-27130129 on Build GRCh38
rs772621502 Chr.2: 27129755-27129756 on Build GRCh38
rs750704153 Chr.2: 27129831-27129831 on Build GRCh38
rs949342199 Chr.2: 27129883-27129883 on Build GRCh38
rs143469823 Chr.2: 27129820-27129820 on Build GRCh38
rs759513048 Chr.2: 27129732-27129732 on Build GRCh38
rs370277773 Chr.2: 27130083-27130083 on Build GRCh38
rs758573221 Chr.2: 27129753-27129753 on Build GRCh38
rs758732729 Chr.2: 27129838-27129838 on Build GRCh38
rs750553276 Chr.2: 27129752-27129752 on Build GRCh38
rs958797668 Chr.2: 27130067-27130067 on Build GRCh38
rs975896291 Chr.2: 27129980-27129980 on Build GRCh38
rs372517067 Chr.2: 27130131-27130131 on Build GRCh38
rs760806677 Chr.2: 27130106-27130106 on Build GRCh38
rs759683962 Chr.2: 27129950-27129950 on Build GRCh38
rs112320078 Chr.2: 27129731-27129731 on Build GRCh38
rs367800329 Chr.2: 27129953-27129953 on Build GRCh38
rs750768265 Chr.2: 27130122-27130122 on Build GRCh38
rs372211196 Chr.2: 27129952-27129952 on Build GRCh38
rs550539325 Chr.2: 27130102-27130102 on Build GRCh38
rs749316316 Chr.2: 27129956-27129963 on Build GRCh38
rs767761176 Chr.2: 27130092-27130092 on Build GRCh38
rs982755297 Chr.2: 27130029-27130030 on Build GRCh38
rs770976721 Chr.2: 27129961-27129962 on Build GRCh38
rs767459354 Chr.2: 27129951-27129951 on Build GRCh38
rs752592315 Chr.2: 27129741-27129741 on Build GRCh38
rs542093633 Chr.2: 27129759-27129759 on Build GRCh38
rs748904019 Chr.2: 27129790-27129790 on Build GRCh38
rs978935299 Chr.2: 27130030-27130030 on Build GRCh38
rs754282181 Chr.2: 27130115-27130115 on Build GRCh38
rs774592256 Chr.2: 27129965-27129966 on Build GRCh38
rs767336752 Chr.2: 27129737-27129737 on Build GRCh38
rs766684467 Chr.2: 27129840-27129840 on Build GRCh38
rs781729723 Chr.2: 27129860-27129860 on Build GRCh38
rs367582724 Chr.2: 27129786-27129786 on Build GRCh38
rs754948236 Chr.2: 27129852-27129852 on Build GRCh38
rs531155473 Chr.2: 27129775-27129775 on Build GRCh38
rs748450927 Chr.2: 27129703-27129703 on Build GRCh38
rs985125572 Chr.2: 27129726-27129726 on Build GRCh38
rs749634291 Chr.2: 27129919-27129919 on Build GRCh38
rs141844750 Chr.2: 27130137-27130137 on Build GRCh38
rs764586880 Chr.2: 27130113-27130113 on Build GRCh38
rs146827545 Chr.2: 27129923-27129923 on Build GRCh38
rs952460824 Chr.2: 27129704-27129704 on Build GRCh38

View Full Product Details