The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004929.3 | NP_004920.1 | 9 - 10 | 787 | 90 |
GenBank mRNA | AK054881.1 | - | 9 - 10 | 785 | 90 |
AK295585.1 | - | 8 - 9 | 506 | 90 | |
AK312491.1 | - | 9 - 10 | 785 | 90 | |
AK315959.1 | - | 8 - 9 | 493 | 90 | |
BC006478.1 | - | 9 - 10 | 775 | 90 | |
BC020864.1 | - | 9 - 10 | 785 | 90 | |
X06661.1 | - | 9 - 10 | 626 | 90 |