The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001194946.1 | NP_001181875.1 | 33 - 34 | 4911 | 67 |
NM_001194947.1 | NP_001181876.1 | 32 - 33 | 4854 | 67 | |
NM_001291157.1 | NP_001278086.1 | 32 - 33 | 4943 | 67 | |
NM_004953.4 | NP_004944.3 | 25 - 26 | 4402 | 67 | |
NM_182917.4 | NP_886553.3 | 31 - 32 | 5038 | 67 | |
NM_198241.2 | NP_937884.1 | 32 - 33 | 4890 | 67 | |
NM_198242.2 | NP_937885.1 | 28 - 29 | 4475 | 67 | |
NM_198244.2 | NP_937887.1 | 29 - 30 | 4652 | 67 | |
GenBank mRNA | AB210013.1 | - | 31 - 32 | 5029 | 67 |
AF104913.1 | - | 29 - 30 | 4514 | 67 | |
AF281070.1 | - | 31 - 32 | 4896 | 67 | |
AK131407.1 | - | 12 - 13 | 1589 | 67 | |
AK226160.1 | - | 32 - 33 | 4824 | 67 | |
AK294567.1 | - | 30 - 31 | 4521 | 67 | |
AK299763.1 | - | 29 - 30 | 4580 | 67 | |
AL832754.1 | - | 28 - 29 | 4364 | 67 | |
AY082886.1 | - | 31 - 32 | 4896 | 67 | |
BC007788.2 | - | 14 - 15 | 1758 | 67 | |
BC082764.1 | - | 26 - 27 | 4013 | 67 | |
BC140892.1 | - | 33 - 34 | 4753 | 67 | |
BC140896.1 | - | 32 - 33 | 4696 | 67 | |
BT007282.1 | - | 14 - 15 | 1757 | 67 | |
BU944064.1 | - | 2 - 3 | 172 | 67 | |
BX647812.1 | - | 32 - 33 | 4803 | 67 | |
D12686.1 | - | 25 - 26 | 4387 | 67 |