Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FUS
Entrez Gene ID
2521
Gene Name
FUS RNA binding protein
Gene Aliases
ALS6, ETM4, FUS1, HNRNPP2, POMP75, TLS
Chromosome Location
Chr.16: 31179444 - 31194871 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170634.1 NP_001164105.1 6 - 7 867 105
NM_001170937.1 NP_001164408.1 6 - 7 858 105
NM_004960.3 NP_004951.1 6 - 7 870 105
XM_011545781.1 XP_011544083.1 6 - 7 836 105
GenBank mRNA AK098774.1 - 6 - 7 838 105
AK290936.1 - 6 - 7 838 105
AK299129.1 - 6 - 7 626 105
BC000402.2 - 6 - 7 833 105
BC002459.1 - 6 - 7 805 105
BC026062.2 - 6 - 7 823 105
BT007131.1 - 5 - 6 765 105
CR456747.1 - 5 - 6 765 105
S62140.1 - 6 - 7 843 105
U36561.1 - 5 - 6 774 105
X71428.1 - 6 - 7 834 105

View Full Product Details