The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005073.3 | NP_005064.1 | 4 - 5 | 321 | 76 |
GenBank mRNA | AB001328.1 | - | 2 - 3 | 340 | 76 |
AF043233.1 | - | 4 - 5 | 300 | 76 | |
AL708816.1 | - | 4 - 5 | 321 | 76 | |
AY634368.1 | - | 4 - 5 | 300 | 76 | |
BC096327.1 | - | 4 - 5 | 275 | 76 | |
BC096328.3 | - | 4 - 5 | 275 | 76 | |
BC096329.3 | - | 4 - 5 | 275 | 76 | |
EU544224.1 | - | 3 - 4 | 244 | 76 | |
U13173.1 | - | 4 - 5 | 300 | 76 | |
U21936.1 | - | 4 - 5 | 300 | 76 |