The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005116.5 | NP_005107.4 | 3 - 4 | 482 | 115 |
NM_203327.1 | NP_976072.1 | 3 - 4 | 500 | 115 | |
XM_011529414.2 | XP_011527716.1 | 3 - 4 | 487 | 115 | |
XM_011529415.2 | XP_011527717.1 | 2 - 3 | 308 | 115 | |
XM_011529416.2 | XP_011527718.1 | 2 - 3 | 617 | 115 | |
XM_011529417.2 | XP_011527719.1 | 2 - 3 | 364 | 115 | |
XM_017028171.1 | XP_016883660.1 | 3 - 4 | 521 | 115 | |
XM_017028172.1 | XP_016883661.1 | 3 - 4 | 525 | 115 | |
XM_017028173.1 | XP_016883662.1 | 3 - 4 | 505 | 115 | |
XM_017028174.1 | XP_016883663.1 | 3 - 4 | 400 | 115 | |
XM_017028176.1 | XP_016883665.1 | 3 - 4 | 496 | 115 | |
XM_017028177.1 | XP_016883666.1 | 3 - 4 | 483 | 115 | |
XM_017028178.1 | XP_016883667.1 | 3 - 4 | 496 | 115 | |
XM_017028179.1 | XP_016883668.1 | 3 - 4 | 483 | 115 | |
XM_017028181.1 | XP_016883670.1 | 3 - 4 | 499 | 115 | |
GenBank mRNA | AF058319.1 | - | 1 - 2 | 107 | 115 |
AF092511.1 | - | 2 - 3 | 403 | 115 | |
AF164142.1 | - | 3 - 4 | 500 | 115 | |
AJ269478.1 | - | 1 - 2 | 107 | 115 | |
AJ292318.1 | - | 2 - 3 | 338 | 115 | |
AK296304.1 | - | 3 - 4 | 490 | 115 | |
AK297253.1 | - | 3 - 4 | 504 | 115 | |
AY380556.1 | - | 1 - 2 | 107 | 115 | |
BC013112.2 | - | 3 - 4 | 471 | 115 | |
D87075.2 | - | 3 - 4 | 482 | 115 | |
EF032501.1 | - | 1 - 2 | 107 | 115 |