Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THOC1
Entrez Gene ID
9984
Gene Name
THO complex 1
Gene Aliases
HPR1, P84, P84N5
Chromosome Location
Chr.18: 214348 - 268059 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK225067.1 - 9 - 10 692 84
AK225709.1 - 9 - 10 680 84
AK314755.1 - 9 - 10 682 84
BC010381.1 - 9 - 10 707 84
L36529.1 - 9 - 10 690 84

Gene Symbol
USP45
Entrez Gene ID
85015
Gene Name
ubiquitin specific peptidase 45
Gene Aliases
-
Chromosome Location
Chr.6: 99432306 - 99515770 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK310643.1 - - 1501 84

View Full Product Details