Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRB10
Entrez Gene ID
2887
Gene Name
growth factor receptor bound protein 10
Gene Aliases
GRB-IR, Grb-10, IRBP, MEG1, RSS
Chromosome Location
Chr.7: 50590063 - 50793462 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001549.2 NP_001001549.1 3 - 4 392 66
NM_001001550.2 NP_001001550.1 3 - 4 472 66
NM_001001555.2 NP_001001555.1 4 - 5 719 66
NM_005311.4 NP_005302.3 3 - 4 392 66
XM_011515304.1 XP_011513606.1 6 - 7 831 66
XM_011515312.2 XP_011513614.1 5 - 6 1119 66
XM_011515316.1 XP_011513618.1 4 - 5 664 66
XM_017012029.1 XP_016867518.1 4 - 5 916 66
XM_017012030.1 XP_016867519.1 3 - 4 850 66
XM_017012031.1 XP_016867520.1 4 - 5 916 66
XM_017012032.1 XP_016867521.1 9 - 10 1293 66
XM_017012033.1 XP_016867522.1 6 - 7 831 66
XM_017012034.1 XP_016867523.1 4 - 5 432 66
XM_017012035.1 XP_016867524.1 10 - 11 2342 66
XM_017012036.1 XP_016867525.1 11 - 12 2127 66
XM_017012037.1 XP_016867526.1 11 - 12 1964 66
XM_017012038.1 XP_016867527.1 10 - 11 1914 66
XM_017012039.1 XP_016867528.1 7 - 8 1161 66
XM_017012040.1 XP_016867529.1 7 - 8 1697 66
XM_017012041.1 XP_016867530.1 9 - 10 1818 66
XM_017012042.1 XP_016867531.1 5 - 6 738 66
XM_017012043.1 XP_016867532.1 6 - 7 928 66
XM_017012044.1 XP_016867533.1 3 - 4 662 66
XM_017012045.1 XP_016867534.1 3 - 4 326 66
XM_017012046.1 XP_016867535.1 7 - 8 1678 66
XM_017012047.1 XP_016867536.1 11 - 12 2526 66
XM_017012048.1 XP_016867537.1 10 - 11 2410 66
XM_017012049.1 XP_016867538.1 11 - 12 2032 66
XM_017012050.1 XP_016867539.1 10 - 11 1982 66
XM_017012051.1 XP_016867540.1 9 - 10 1886 66
XM_017012052.1 XP_016867541.1 9 - 10 2226 66
XM_017012053.1 XP_016867542.1 10 - 11 1848 66
XM_017012054.1 XP_016867543.1 9 - 10 1798 66
XM_017012055.1 XP_016867544.1 6 - 7 1581 66
XM_017012056.1 XP_016867545.1 5 - 6 2413 66
XM_017012057.1 XP_016867546.1 8 - 9 1702 66
XM_017012058.1 XP_016867547.1 4 - 5 573 66
XM_017012059.1 XP_016867548.1 4 - 5 695 66
XM_017012060.1 XP_016867549.1 12 - 13 2659 66
XM_017012061.1 XP_016867550.1 3 - 4 525 66
XM_017012062.1 XP_016867551.1 2 - 3 286 66
XM_017012063.1 XP_016867552.1 11 - 12 2526 66
XM_017012064.1 XP_016867553.1 10 - 11 2410 66
XM_017012065.1 XP_016867554.1 8 - 9 1862 66
XM_017012066.1 XP_016867555.1 6 - 7 2221 66
XM_017012067.1 XP_016867556.1 6 - 7 724 66
XM_017012068.1 XP_016867557.1 6 - 7 1590 66
GenBank mRNA AB000731.1 - 3 - 4 367 66
AF000017.1 - 3 - 4 361 66
AF001534.1 - 3 - 4 392 66
AJ271366.1 - 6 - 7 831 66
BC024285.1 - 4 - 5 496 66
BX649028.1 - 3 - 4 452 66
D86962.1 - 5 - 6 1124 66
U34355.1 - 3 - 4 392 66
U66065.1 - 3 - 4 472 66
U69276.1 - 3 - 4 474 66

View Full Product Details