The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005383.2 | NP_005374.2 | 1 - 2 | 202 | 113 |
GenBank mRNA | BC069151.1 | - | 1 - 2 | 202 | 113 |
BC107053.2 | - | 1 - 2 | 202 | 113 |