The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284209.1 | NP_001271138.1 | 1 - 2 | 79 | 57 |
NM_001284210.1 | NP_001271139.1 | 1 - 2 | 79 | 57 | |
NM_001284211.1 | NP_001271140.1 | 1 - 2 | 79 | 57 | |
NM_005466.3 | NP_005457.2 | 1 - 2 | 79 | 57 | |
GenBank mRNA | AF074723.1 | - | 1 - 2 | 50 | 57 |
AK222587.1 | - | 1 - 2 | 55 | 57 | |
AK223346.1 | - | 1 - 2 | 44 | 57 | |
AK290647.1 | - | 1 - 2 | 44 | 57 | |
AK300417.1 | - | 1 - 2 | 44 | 57 | |
AK300460.1 | - | 1 - 2 | 44 | 57 | |
AK304361.1 | - | 1 - 2 | 44 | 57 | |
BC004106.2 | - | 1 - 2 | 53 | 57 | |
HY064772.1 | - | 1 - 2 | 80 | 57 | |
U78082.1 | - | 1 - 2 | 76 | 57 |