The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206741.1 | NP_001193670.1 | 5 - 6 | 669 | 77 |
NM_005525.3 | NP_005516.1 | 4 - 5 | 667 | 77 | |
NM_181755.2 | NP_861420.1 | 5 - 6 | 647 | 77 | |
GenBank mRNA | AK313973.1 | - | 4 - 5 | 609 | 77 |
BC012593.1 | - | 4 - 5 | 616 | 77 | |
BX345837.2 | - | 4 - 5 | 580 | 77 | |
KJ534869.1 | - | 4 - 5 | 515 | 77 |