The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300804.1 | NP_001287733.1 | 5 - 6 | 831 | 73 |
NM_001300805.1 | NP_001287734.1 | 3 - 4 | 511 | 73 | |
NM_001300806.1 | NP_001287735.1 | 3 - 4 | 780 | 73 | |
NM_005586.3 | NP_005577.1 | 4 - 5 | 694 | 73 | |
XM_005249117.3 | XP_005249174.1 | 4 - 5 | 697 | 73 | |
XM_011514625.2 | XP_011512927.1 | 5 - 6 | 982 | 73 | |
XM_011514626.2 | XP_011512928.1 | 4 - 5 | 805 | 73 | |
XM_017010867.1 | XP_016866356.1 | 3 - 4 | 1353 | 73 | |
GenBank mRNA | AK290735.1 | - | 3 - 4 | 778 | 73 |
BC007836.2 | - | 4 - 5 | 643 | 73 | |
BI824391.1 | - | 3 - 4 | 491 | 73 | |
BX397111.2 | - | 5 - 6 | 814 | 73 | |
U78313.1 | - | 4 - 5 | 636 | 73 |