The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001185056.1 | NP_001171985.1 | 2 - 3 | 268 | 60 |
NM_005602.5 | NP_005593.2 | 2 - 3 | 598 | 60 | |
GenBank mRNA | AJ245901.1 | - | 2 - 3 | 396 | 60 |
AK294087.1 | - | 2 - 3 | 239 | 60 | |
AK312923.1 | - | 2 - 3 | 595 | 60 | |
BC013577.1 | - | 2 - 3 | 586 | 60 | |
BT019402.1 | - | 2 - 3 | 396 | 60 | |
BT019403.1 | - | 2 - 3 | 396 | 60 | |
DA258090.1 | - | 2 - 3 | 268 | 60 |