Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69072929-69073476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANTXR1
Entrez Gene ID
84168
Gene Name
anthrax toxin receptor 1
Gene Aliases
ATR, GAPO, TEM8
UniGene
Hs.165859
Gene Chromosome Location
Chr.2: 69013144-69249327 on Build GRCh38
Transcripts
NM_018153.3 NM_032208.2 NM_053034.2 XM_017005075.1 XM_017005076.1 XM_017005077.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4095272 c.468T>A p.D156E Chr.2: 69073077-69073077 on Build GRCh38
6325060 c.413-1G>A p.? Chr.2: 69073021-69073021 on Build GRCh38
4416533 c.492+10G>A p.? Chr.2: 69073111-69073111 on Build GRCh38
6269166 c.492+10G>T p.? Chr.2: 69073111-69073111 on Build GRCh38
5422425 c.482C>T p.S161L Chr.2: 69073091-69073091 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs948718096 Chr.2: 69073329-69073329 on Build GRCh38
rs189998937 Chr.2: 69073302-69073302 on Build GRCh38
rs764736320 Chr.2: 69073152-69073152 on Build GRCh38
rs370376652 Chr.2: 69073030-69073030 on Build GRCh38
rs538123354 Chr.2: 69072955-69072958 on Build GRCh38
rs376767455 Chr.2: 69073230-69073230 on Build GRCh38
rs775780859 Chr.2: 69072983-69072983 on Build GRCh38
rs67110959 Chr.2: 69072957-69072959 on Build GRCh38
rs1000710232 Chr.2: 69072986-69072986 on Build GRCh38
rs942791300 Chr.2: 69073196-69073196 on Build GRCh38
rs1025760615 Chr.2: 69073170-69073170 on Build GRCh38
rs557289749 Chr.2: 69073194-69073194 on Build GRCh38
rs767288834 Chr.2: 69072988-69072988 on Build GRCh38
rs766350438 Chr.2: 69073328-69073328 on Build GRCh38
rs923859133 Chr.2: 69073273-69073273 on Build GRCh38
rs200355895 Chr.2: 69073080-69073080 on Build GRCh38
rs752336491 Chr.2: 69073052-69073052 on Build GRCh38
rs867503360 Chr.2: 69072956-69072959 on Build GRCh38
rs563626271 Chr.2: 69073403-69073403 on Build GRCh38
rs759323987 Chr.2: 69073430-69073430 on Build GRCh38
rs753396370 Chr.2: 69073035-69073035 on Build GRCh38
rs772943487 Chr.2: 69073133-69073133 on Build GRCh38
rs112979831 Chr.2: 69073322-69073322 on Build GRCh38
rs758980852 Chr.2: 69073145-69073145 on Build GRCh38
rs548701918 Chr.2: 69072961-69072961 on Build GRCh38
rs953132744 Chr.2: 69073238-69073239 on Build GRCh38
rs1046062850 Chr.2: 69073297-69073297 on Build GRCh38
rs201593539 Chr.2: 69072994-69072994 on Build GRCh38
rs760441318 Chr.2: 69073136-69073136 on Build GRCh38
rs559562299 Chr.2: 69073234-69073234 on Build GRCh38
rs771654357 Chr.2: 69073255-69073255 on Build GRCh38
rs769908738 Chr.2: 69073117-69073117 on Build GRCh38
rs79912752 Chr.2: 69073429-69073429 on Build GRCh38
rs76355575 Chr.2: 69073177-69073177 on Build GRCh38
rs984698183 Chr.2: 69073190-69073190 on Build GRCh38
rs529509144 Chr.2: 69073409-69073409 on Build GRCh38
rs370064775 Chr.2: 69072954-69072955 on Build GRCh38
rs181468623 Chr.2: 69073427-69073427 on Build GRCh38
rs760702171 Chr.2: 69073311-69073311 on Build GRCh38
rs34626010 Chr.2: 69073198-69073199 on Build GRCh38
rs952847746 Chr.2: 69073185-69073185 on Build GRCh38
rs763171813 Chr.2: 69073120-69073120 on Build GRCh38
rs778487700 Chr.2: 69073039-69073039 on Build GRCh38
rs113138542 Chr.2: 69073142-69073142 on Build GRCh38
rs60334682 Chr.2: 69072957-69072957 on Build GRCh38
rs4854537 Chr.2: 69073332-69073332 on Build GRCh38
rs1029903938 Chr.2: 69073193-69073193 on Build GRCh38
rs10202939 Chr.2: 69073111-69073111 on Build GRCh38
rs936467659 Chr.2: 69073276-69073276 on Build GRCh38
rs771612099 Chr.2: 69073089-69073089 on Build GRCh38
rs761574644 Chr.2: 69072987-69072987 on Build GRCh38
rs754480769 Chr.2: 69073038-69073038 on Build GRCh38
rs77687153 Chr.2: 69073059-69073059 on Build GRCh38
rs573908971 Chr.2: 69073205-69073205 on Build GRCh38
rs67323679 Chr.2: 69072959-69072961 on Build GRCh38
rs57008189 Chr.2: 69072955-69072955 on Build GRCh38
rs771888374 Chr.2: 69073129-69073129 on Build GRCh38
rs765898875 Chr.2: 69073033-69073033 on Build GRCh38
rs750236224 Chr.2: 69072956-69072957 on Build GRCh38
rs766345099 Chr.2: 69073139-69073139 on Build GRCh38
rs1012433906 Chr.2: 69073289-69073289 on Build GRCh38
rs757240018 Chr.2: 69073110-69073110 on Build GRCh38
rs922618983 Chr.2: 69073225-69073225 on Build GRCh38
rs1025091698 Chr.2: 69073122-69073122 on Build GRCh38
rs1054043567 Chr.2: 69073278-69073278 on Build GRCh38
rs533172761 Chr.2: 69073305-69073305 on Build GRCh38
rs768226090 Chr.2: 69073071-69073071 on Build GRCh38
rs550479175 Chr.2: 69073175-69073175 on Build GRCh38
rs1045734447 Chr.2: 69073330-69073330 on Build GRCh38
rs386647050 Chr.2: 69072955-69072961 on Build GRCh38
rs111489161 Chr.2: 69073266-69073266 on Build GRCh38
rs184852244 Chr.2: 69073288-69073288 on Build GRCh38
rs113688651 Chr.2: 69073249-69073250 on Build GRCh38
rs976765552 Chr.2: 69073217-69073217 on Build GRCh38
rs756008471 Chr.2: 69073029-69073029 on Build GRCh38

View Full Product Details