The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005715.2 | NP_005706.1 | 5 - 6 | 974 | 121 |
GenBank mRNA | AB020316.1 | - | 5 - 6 | 781 | 121 |
AK292922.1 | - | 5 - 6 | 722 | 121 | |
AK315320.1 | - | 5 - 6 | 682 | 121 | |
BC093668.1 | - | 5 - 6 | 726 | 121 | |
BC093694.1 | - | 5 - 6 | 726 | 121 |