Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70677677-70678171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADD2
Entrez Gene ID
119
Gene Name
adducin 2
Gene Aliases
ADDB
UniGene
Hs.188528
Gene Chromosome Location
Chr.2: 70656784-70768243 on Build GRCh38
Transcripts
NM_001185054.1 NM_001185055.1 NM_001617.3 NM_017482.3 NM_017488.3 XM_011532502.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1409182 c.1454T>G p.F485C Chr.2: 70677807-70677807 on Build GRCh38
5911576 c.1489G>A p.E497K Chr.2: 70677772-70677772 on Build GRCh38
7106888 c.1453G>A p.E485K Chr.2: 70677856-70677856 on Build GRCh38
4659990 c.1530A>C p.E510D Chr.2: 70677779-70677779 on Build GRCh38
4659988 c.1482A>C p.E494D Chr.2: 70677779-70677779 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782218257 Chr.2: 70677901-70677901 on Build GRCh38
rs782632099 Chr.2: 70677909-70677909 on Build GRCh38
rs1040979570 Chr.2: 70677941-70677941 on Build GRCh38
rs782163938 Chr.2: 70677720-70677720 on Build GRCh38
rs894155512 Chr.2: 70678071-70678071 on Build GRCh38
rs374495574 Chr.2: 70677872-70677872 on Build GRCh38
rs781951897 Chr.2: 70677834-70677834 on Build GRCh38
rs781877025 Chr.2: 70677758-70677758 on Build GRCh38
rs145104392 Chr.2: 70678098-70678098 on Build GRCh38
rs782723909 Chr.2: 70677874-70677874 on Build GRCh38
rs79548563 Chr.2: 70678097-70678097 on Build GRCh38
rs1012901745 Chr.2: 70678075-70678075 on Build GRCh38
rs782693854 Chr.2: 70677908-70677908 on Build GRCh38
rs782205659 Chr.2: 70677713-70677713 on Build GRCh38
rs782818996 Chr.2: 70677889-70677889 on Build GRCh38
rs375540457 Chr.2: 70677709-70677709 on Build GRCh38
rs904240799 Chr.2: 70678001-70678001 on Build GRCh38
rs782169069 Chr.2: 70677918-70677918 on Build GRCh38
rs782479382 Chr.2: 70677812-70677812 on Build GRCh38
rs782096302 Chr.2: 70677709-70677710 on Build GRCh38
rs150426440 Chr.2: 70677828-70677828 on Build GRCh38
rs781832361 Chr.2: 70677803-70677803 on Build GRCh38
rs782425601 Chr.2: 70677752-70677752 on Build GRCh38
rs970900790 Chr.2: 70677847-70677847 on Build GRCh38
rs2902577 Chr.2: 70677967-70677967 on Build GRCh38
rs781949931 Chr.2: 70677919-70677919 on Build GRCh38
rs781913377 Chr.2: 70677868-70677868 on Build GRCh38
rs555651168 Chr.2: 70678044-70678044 on Build GRCh38
rs192095501 Chr.2: 70678101-70678101 on Build GRCh38
rs782522514 Chr.2: 70677908-70677908 on Build GRCh38
rs781922267 Chr.2: 70677723-70677723 on Build GRCh38
rs782492778 Chr.2: 70677883-70677883 on Build GRCh38
rs782565347 Chr.2: 70677795-70677795 on Build GRCh38
rs377431981 Chr.2: 70677711-70677711 on Build GRCh38
rs782758494 Chr.2: 70677924-70677924 on Build GRCh38
rs188058882 Chr.2: 70677863-70677863 on Build GRCh38
rs779365593 Chr.2: 70677993-70677993 on Build GRCh38
rs782390307 Chr.2: 70677714-70677714 on Build GRCh38
rs781869077 Chr.2: 70677875-70677875 on Build GRCh38
rs1000304086 Chr.2: 70677717-70677717 on Build GRCh38
rs918711949 Chr.2: 70677882-70677882 on Build GRCh38
rs781944749 Chr.2: 70678119-70678119 on Build GRCh38
rs781930613 Chr.2: 70677823-70677823 on Build GRCh38
rs782130731 Chr.2: 70677920-70677920 on Build GRCh38
rs200444585 Chr.2: 70677710-70677710 on Build GRCh38
rs539217357 Chr.2: 70678005-70678005 on Build GRCh38
rs782818465 Chr.2: 70677775-70677775 on Build GRCh38
rs945885720 Chr.2: 70677971-70677971 on Build GRCh38
rs73937001 Chr.2: 70677897-70677897 on Build GRCh38
rs782409297 Chr.2: 70677913-70677913 on Build GRCh38
rs782294118 Chr.2: 70677910-70677910 on Build GRCh38
rs527878592 Chr.2: 70677790-70677790 on Build GRCh38
rs34884423 Chr.2: 70677833-70677833 on Build GRCh38
rs781946199 Chr.2: 70677917-70677917 on Build GRCh38
rs1041485663 Chr.2: 70677990-70677990 on Build GRCh38
rs13306101 Chr.2: 70677737-70677737 on Build GRCh38
rs782414691 Chr.2: 70677826-70677826 on Build GRCh38
rs781894360 Chr.2: 70677781-70677781 on Build GRCh38
rs991105341 Chr.2: 70677817-70677817 on Build GRCh38
rs782263489 Chr.2: 70677835-70677835 on Build GRCh38
rs781883883 Chr.2: 70677891-70677891 on Build GRCh38
rs565926773 Chr.2: 70678064-70678064 on Build GRCh38
rs782329547 Chr.2: 70677722-70677722 on Build GRCh38
rs782384294 Chr.2: 70677840-70677840 on Build GRCh38
rs782037969 Chr.2: 70677849-70677849 on Build GRCh38
rs782151348 Chr.2: 70677862-70677862 on Build GRCh38
rs782437715 Chr.2: 70677896-70677896 on Build GRCh38
rs781890019 Chr.2: 70677911-70677911 on Build GRCh38
rs781982582 Chr.2: 70677716-70677716 on Build GRCh38
rs148922479 Chr.2: 70678103-70678103 on Build GRCh38
rs550494762 Chr.2: 70677904-70677904 on Build GRCh38
rs199802003 Chr.2: 70677824-70677824 on Build GRCh38
rs570030264 Chr.2: 70677950-70677950 on Build GRCh38
rs112879637 Chr.2: 70677759-70677759 on Build GRCh38
rs782183958 Chr.2: 70677830-70677830 on Build GRCh38
rs543472154 Chr.2: 70678110-70678110 on Build GRCh38

View Full Product Details