The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102398.2 | NP_001095868.1 | 1 - 2 | 159 | 87 |
NM_001297620.1 | NP_001284549.1 | 1 - 2 | 159 | 87 | |
NM_005826.4 | NP_005817.1 | 1 - 2 | 159 | 87 | |
XM_011540475.2 | XP_011538777.1 | 1 - 2 | 445 | 87 | |
XM_017000010.1 | XP_016855499.1 | 1 - 2 | 96 | 87 | |
GenBank mRNA | AF000364.1 | - | 1 - 2 | 57 | 87 |
AK297382.1 | - | 1 - 2 | 76 | 87 | |
AK313142.1 | - | 1 - 2 | 107 | 87 | |
BC001449.2 | - | 1 - 2 | 105 | 87 |