Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74562127-74562635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
M1AP
Entrez Gene ID
130951
Gene Name
meiosis 1 associated protein
Gene Aliases
C2orf65, D6Mm5e, SPATA37
UniGene
Hs.348645
Gene Chromosome Location
Chr.2: 74557640-74648338 on Build GRCh38
Transcripts
NM_001281296.1 NM_001321739.1 NM_138804.4 XM_005264152.3 XM_006711946.3 XM_011532548.2 XM_011532549.2 XM_011532550.2 XM_011532551.2 XM_011532552.2 XM_011532553.2 XM_011532554.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7239397 c.1075-8T>C p.? Chr.2: 74562431-74562431 on Build GRCh38
4095832 c.1256A>C p.H419P Chr.2: 74562242-74562242 on Build GRCh38
4757576 c.1174T>G p.S392A Chr.2: 74562324-74562324 on Build GRCh38
1409616 c.1222C>A p.P408T Chr.2: 74562276-74562276 on Build GRCh38
7227180 c.1111C>A p.P371T Chr.2: 74562387-74562387 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767015429 Chr.2: 74562457-74562457 on Build GRCh38
rs754994144 Chr.2: 74562383-74562383 on Build GRCh38
rs976932817 Chr.2: 74562366-74562366 on Build GRCh38
rs539290626 Chr.2: 74562389-74562389 on Build GRCh38
rs756858175 Chr.2: 74562333-74562333 on Build GRCh38
rs375676422 Chr.2: 74562434-74562434 on Build GRCh38
rs775925196 Chr.2: 74562409-74562409 on Build GRCh38
rs200523774 Chr.2: 74562380-74562380 on Build GRCh38
rs941290363 Chr.2: 74562472-74562472 on Build GRCh38
rs750188912 Chr.2: 74562350-74562350 on Build GRCh38
rs1035523702 Chr.2: 74562272-74562272 on Build GRCh38
rs35386699 Chr.2: 74562504-74562505 on Build GRCh38
rs777102056 Chr.2: 74562177-74562177 on Build GRCh38
rs766256428 Chr.2: 74562467-74562467 on Build GRCh38
rs141186317 Chr.2: 74562382-74562382 on Build GRCh38
rs952384533 Chr.2: 74562337-74562337 on Build GRCh38
rs1050839071 Chr.2: 74562204-74562204 on Build GRCh38
rs765800421 Chr.2: 74562178-74562178 on Build GRCh38
rs560457393 Chr.2: 74562307-74562307 on Build GRCh38
rs748173359 Chr.2: 74562291-74562291 on Build GRCh38
rs781120910 Chr.2: 74562223-74562223 on Build GRCh38
rs368526345 Chr.2: 74562451-74562451 on Build GRCh38
rs747098542 Chr.2: 74562596-74562596 on Build GRCh38
rs895213334 Chr.2: 74562547-74562547 on Build GRCh38
rs1031235139 Chr.2: 74562588-74562588 on Build GRCh38
rs772004777 Chr.2: 74562224-74562226 on Build GRCh38
rs776853780 Chr.2: 74562318-74562318 on Build GRCh38
rs200133274 Chr.2: 74562385-74562385 on Build GRCh38
rs561022917 Chr.2: 74562579-74562579 on Build GRCh38
rs997837856 Chr.2: 74562610-74562610 on Build GRCh38
rs779814442 Chr.2: 74562379-74562380 on Build GRCh38
rs114430238 Chr.2: 74562509-74562509 on Build GRCh38
rs767262447 Chr.2: 74562343-74562343 on Build GRCh38
rs780821512 Chr.2: 74562243-74562243 on Build GRCh38
rs757024741 Chr.2: 74562235-74562235 on Build GRCh38
rs533977848 Chr.2: 74562232-74562232 on Build GRCh38
rs879088529 Chr.2: 74562431-74562431 on Build GRCh38
rs202079854 Chr.2: 74562336-74562336 on Build GRCh38
rs765673853 Chr.2: 74562323-74562323 on Build GRCh38
rs771044914 Chr.2: 74562386-74562386 on Build GRCh38
rs780099442 Chr.2: 74562379-74562379 on Build GRCh38
rs11126435 Chr.2: 74562573-74562573 on Build GRCh38
rs186424282 Chr.2: 74562331-74562331 on Build GRCh38
rs538415282 Chr.2: 74562377-74562377 on Build GRCh38
rs889098780 Chr.2: 74562207-74562207 on Build GRCh38
rs143680043 Chr.2: 74562222-74562222 on Build GRCh38
rs761062952 Chr.2: 74562172-74562172 on Build GRCh38
rs377032064 Chr.2: 74562202-74562202 on Build GRCh38
rs372490474 Chr.2: 74562464-74562464 on Build GRCh38
rs745610891 Chr.2: 74562265-74562265 on Build GRCh38
rs904023429 Chr.2: 74562568-74562568 on Build GRCh38
rs895348981 Chr.2: 74562254-74562254 on Build GRCh38
rs771223238 Chr.2: 74562302-74562302 on Build GRCh38
rs773723427 Chr.2: 74562437-74562437 on Build GRCh38
rs752189876 Chr.2: 74562180-74562180 on Build GRCh38
rs370086618 Chr.2: 74562301-74562301 on Build GRCh38
rs367719199 Chr.2: 74562292-74562292 on Build GRCh38
rs770171453 Chr.2: 74562392-74562392 on Build GRCh38
rs1056531366 Chr.2: 74562241-74562241 on Build GRCh38
rs779960080 Chr.2: 74562465-74562465 on Build GRCh38
rs116670856 Chr.2: 74562566-74562566 on Build GRCh38
rs763343614 Chr.2: 74562412-74562412 on Build GRCh38
rs748285504 Chr.2: 74562493-74562493 on Build GRCh38
rs745727688 Chr.2: 74562249-74562249 on Build GRCh38
rs751126701 Chr.2: 74562332-74562332 on Build GRCh38
rs368854401 Chr.2: 74562262-74562262 on Build GRCh38
rs759950731 Chr.2: 74562319-74562319 on Build GRCh38
rs745662064 Chr.2: 74562172-74562172 on Build GRCh38
rs773436081 Chr.2: 74562293-74562293 on Build GRCh38
rs79405350 Chr.2: 74562175-74562175 on Build GRCh38
rs780156712 Chr.2: 74562288-74562288 on Build GRCh38
rs1012373750 Chr.2: 74562561-74562561 on Build GRCh38
rs939357123 Chr.2: 74562504-74562504 on Build GRCh38
rs775976762 Chr.2: 74562325-74562325 on Build GRCh38
rs1036946569 Chr.2: 74562492-74562492 on Build GRCh38

View Full Product Details