Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74576322-74576821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
M1AP
Entrez Gene ID
130951
Gene Name
meiosis 1 associated protein
Gene Aliases
C2orf65, D6Mm5e, SPATA37
UniGene
Hs.348645
Gene Chromosome Location
Chr.2: 74557640-74648338 on Build GRCh38
Transcripts
NM_001281295.1 NM_001281296.1 NM_001321739.1 NM_138804.4 XM_005264152.3 XM_006711946.3 XM_011532548.2 XM_011532549.2 XM_011532550.2 XM_011532551.2 XM_011532552.2 XM_011532553.2 XM_011532554.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1023080 c.862G>T p.G288* Chr.2: 74576526-74576526 on Build GRCh38
6900448 c.793G>A p.E265K Chr.2: 74576595-74576595 on Build GRCh38
5862887 c.820G>T p.A274S Chr.2: 74576568-74576568 on Build GRCh38
443240 c.844A>T p.R282* Chr.2: 74576544-74576544 on Build GRCh38
1163265 c.917_918insA p.L307fs*10 Chr.2: 74576470-74576471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367990203 Chr.2: 74576553-74576553 on Build GRCh38
rs748435404 Chr.2: 74576660-74576660 on Build GRCh38
rs112300450 Chr.2: 74576348-74576348 on Build GRCh38
rs1056551335 Chr.2: 74576462-74576462 on Build GRCh38
rs755613339 Chr.2: 74576637-74576637 on Build GRCh38
rs142319651 Chr.2: 74576549-74576549 on Build GRCh38
rs758836998 Chr.2: 74576540-74576540 on Build GRCh38
rs753905899 Chr.2: 74576523-74576523 on Build GRCh38
rs878930806 Chr.2: 74576729-74576729 on Build GRCh38
rs887578675 Chr.2: 74576767-74576767 on Build GRCh38
rs755205766 Chr.2: 74576419-74576419 on Build GRCh38
rs746452484 Chr.2: 74576456-74576456 on Build GRCh38
rs562596278 Chr.2: 74576533-74576533 on Build GRCh38
rs201666042 Chr.2: 74576444-74576444 on Build GRCh38
rs267599461 Chr.2: 74576500-74576500 on Build GRCh38
rs755225252 Chr.2: 74576527-74576527 on Build GRCh38
rs752008624 Chr.2: 74576614-74576614 on Build GRCh38
rs1049148225 Chr.2: 74576748-74576748 on Build GRCh38
rs373451233 Chr.2: 74576633-74576633 on Build GRCh38
rs960105842 Chr.2: 74576364-74576364 on Build GRCh38
rs751727560 Chr.2: 74576495-74576500 on Build GRCh38
rs138006701 Chr.2: 74576486-74576486 on Build GRCh38
rs754112970 Chr.2: 74576418-74576418 on Build GRCh38
rs569218987 Chr.2: 74576796-74576796 on Build GRCh38
rs752816889 Chr.2: 74576561-74576577 on Build GRCh38
rs948111190 Chr.2: 74576723-74576723 on Build GRCh38
rs150497323 Chr.2: 74576592-74576592 on Build GRCh38
rs201780018 Chr.2: 74576617-74576617 on Build GRCh38
rs138394848 Chr.2: 74576654-74576654 on Build GRCh38
rs550638946 Chr.2: 74576714-74576714 on Build GRCh38
rs143537845 Chr.2: 74576578-74576578 on Build GRCh38
rs62147439 Chr.2: 74576399-74576399 on Build GRCh38
rs770596190 Chr.2: 74576457-74576457 on Build GRCh38
rs745433133 Chr.2: 74576554-74576554 on Build GRCh38
rs761708409 Chr.2: 74576570-74576570 on Build GRCh38
rs10170219 Chr.2: 74576390-74576390 on Build GRCh38
rs544319090 Chr.2: 74576501-74576501 on Build GRCh38
rs924230416 Chr.2: 74576652-74576652 on Build GRCh38
rs184357128 Chr.2: 74576381-74576381 on Build GRCh38
rs759545535 Chr.2: 74576580-74576580 on Build GRCh38
rs772253961 Chr.2: 74576712-74576712 on Build GRCh38
rs759729863 Chr.2: 74576521-74576522 on Build GRCh38
rs764527856 Chr.2: 74576598-74576598 on Build GRCh38
rs760700924 Chr.2: 74576503-74576503 on Build GRCh38
rs757259912 Chr.2: 74576464-74576464 on Build GRCh38
rs778437288 Chr.2: 74576447-74576447 on Build GRCh38
rs573428972 Chr.2: 74576431-74576431 on Build GRCh38
rs766758402 Chr.2: 74576478-74576479 on Build GRCh38
rs753051076 Chr.2: 74576535-74576535 on Build GRCh38
rs776760571 Chr.2: 74576579-74576579 on Build GRCh38
rs34267938 Chr.2: 74576495-74576496 on Build GRCh38
rs10182717 Chr.2: 74576636-74576636 on Build GRCh38
rs142260852 Chr.2: 74576566-74576566 on Build GRCh38
rs374484414 Chr.2: 74576665-74576665 on Build GRCh38
rs200361166 Chr.2: 74576569-74576569 on Build GRCh38
rs113893192 Chr.2: 74576653-74576653 on Build GRCh38
rs145804668 Chr.2: 74576485-74576485 on Build GRCh38
rs200989583 Chr.2: 74576546-74576546 on Build GRCh38
rs779711121 Chr.2: 74576638-74576638 on Build GRCh38
rs766461019 Chr.2: 74576511-74576511 on Build GRCh38
rs779580170 Chr.2: 74576565-74576565 on Build GRCh38
rs895161675 Chr.2: 74576716-74576716 on Build GRCh38
rs776094291 Chr.2: 74576468-74576468 on Build GRCh38
rs376663636 Chr.2: 74576482-74576482 on Build GRCh38
rs779306793 Chr.2: 74576426-74576426 on Build GRCh38
rs768465839 Chr.2: 74576568-74576568 on Build GRCh38
rs149272394 Chr.2: 74576591-74576591 on Build GRCh38
rs767669802 Chr.2: 74576557-74576557 on Build GRCh38
rs745508636 Chr.2: 74576480-74576480 on Build GRCh38
rs150145101 Chr.2: 74576582-74576582 on Build GRCh38

View Full Product Details