Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96848365-96848863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD39
Entrez Gene ID
51239
Gene Name
ankyrin repeat domain 39
Gene Aliases
-
UniGene
Hs.709507
Gene Chromosome Location
Chr.2: 96847985-96858019 on Build GRCh38
Transcripts
NM_016466.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7282782 c.442_443insA p.L148fs*22 Chr.2: 96848410-96848411 on Build GRCh38
4096445 c.478C>T p.R160W Chr.2: 96848375-96848375 on Build GRCh38
1307115 c.415G>A p.E139K Chr.2: 96848438-96848438 on Build GRCh38
477778 c.488A>G p.K163R Chr.2: 96848365-96848365 on Build GRCh38
2825866 c.467T>C p.L156P Chr.2: 96848386-96848386 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534192996 Chr.2: 96848744-96848744 on Build GRCh38
rs769004469 Chr.2: 96848745-96848745 on Build GRCh38
rs911353392 Chr.2: 96848831-96848831 on Build GRCh38
rs748672225 Chr.2: 96848813-96848814 on Build GRCh38
rs775403383 Chr.2: 96848391-96848391 on Build GRCh38
rs987374271 Chr.2: 96848595-96848595 on Build GRCh38
rs369423598 Chr.2: 96848482-96848482 on Build GRCh38
rs942432765 Chr.2: 96848603-96848603 on Build GRCh38
rs1025641186 Chr.2: 96848796-96848796 on Build GRCh38
rs763051737 Chr.2: 96848393-96848393 on Build GRCh38
rs1023492570 Chr.2: 96848514-96848514 on Build GRCh38
rs777412788 Chr.2: 96848502-96848502 on Build GRCh38
rs762188362 Chr.2: 96848413-96848413 on Build GRCh38
rs1001994741 Chr.2: 96848770-96848770 on Build GRCh38
rs546669250 Chr.2: 96848597-96848597 on Build GRCh38
rs761726697 Chr.2: 96848404-96848406 on Build GRCh38
rs1043498821 Chr.2: 96848646-96848646 on Build GRCh38
rs781392554 Chr.2: 96848459-96848459 on Build GRCh38
rs111987165 Chr.2: 96848648-96848648 on Build GRCh38
rs962696242 Chr.2: 96848551-96848551 on Build GRCh38
rs1043769482 Chr.2: 96848405-96848405 on Build GRCh38
rs766102463 Chr.2: 96848485-96848485 on Build GRCh38
rs1038402657 Chr.2: 96848767-96848767 on Build GRCh38
rs778401253 Chr.2: 96848447-96848447 on Build GRCh38
rs375112201 Chr.2: 96848489-96848489 on Build GRCh38
rs567180236 Chr.2: 96848518-96848518 on Build GRCh38
rs924918301 Chr.2: 96848685-96848685 on Build GRCh38
rs934125020 Chr.2: 96848592-96848592 on Build GRCh38
rs754429055 Chr.2: 96848436-96848436 on Build GRCh38
rs754978297 Chr.2: 96848490-96848490 on Build GRCh38
rs1034942426 Chr.2: 96848568-96848568 on Build GRCh38
rs547928954 Chr.2: 96848637-96848637 on Build GRCh38
rs567608757 Chr.2: 96848503-96848503 on Build GRCh38
rs918839990 Chr.2: 96848560-96848560 on Build GRCh38
rs770671896 Chr.2: 96848601-96848601 on Build GRCh38
rs180811750 Chr.2: 96848478-96848478 on Build GRCh38
rs934170873 Chr.2: 96848743-96848743 on Build GRCh38
rs774102817 Chr.2: 96848638-96848638 on Build GRCh38
rs956568963 Chr.2: 96848587-96848587 on Build GRCh38
rs940249738 Chr.2: 96848756-96848756 on Build GRCh38
rs946433941 Chr.2: 96848473-96848473 on Build GRCh38
rs1017035711 Chr.2: 96848782-96848782 on Build GRCh38
rs377600119 Chr.2: 96848799-96848799 on Build GRCh38
rs753739984 Chr.2: 96848432-96848432 on Build GRCh38
rs536699823 Chr.2: 96848509-96848509 on Build GRCh38
rs962646096 Chr.2: 96848784-96848784 on Build GRCh38
rs1052318632 Chr.2: 96848694-96848694 on Build GRCh38
rs147146215 Chr.2: 96848691-96848691 on Build GRCh38
rs773843058 Chr.2: 96848565-96848565 on Build GRCh38
rs914999145 Chr.2: 96848463-96848463 on Build GRCh38
rs750281190 Chr.2: 96848424-96848424 on Build GRCh38
rs898473467 Chr.2: 96848795-96848795 on Build GRCh38
rs975547717 Chr.2: 96848729-96848729 on Build GRCh38
rs773032708 Chr.2: 96848478-96848478 on Build GRCh38
rs987101702 Chr.2: 96848817-96848817 on Build GRCh38
rs531650049 Chr.2: 96848820-96848821 on Build GRCh38
rs145738875 Chr.2: 96848426-96848426 on Build GRCh38
rs767403087 Chr.2: 96848421-96848421 on Build GRCh38
rs896396741 Chr.2: 96848721-96848721 on Build GRCh38
rs771770206 Chr.2: 96848458-96848458 on Build GRCh38
rs759950256 Chr.2: 96848386-96848386 on Build GRCh38
rs905053738 Chr.2: 96848752-96848752 on Build GRCh38
rs367801086 Chr.2: 96848411-96848411 on Build GRCh38
rs200007178 Chr.2: 96848483-96848483 on Build GRCh38
rs779999285 Chr.2: 96848427-96848427 on Build GRCh38
rs528356666 Chr.2: 96848454-96848454 on Build GRCh38
rs371933512 Chr.2: 96848474-96848474 on Build GRCh38
rs770385380 Chr.2: 96848389-96848389 on Build GRCh38
rs749001442 Chr.2: 96848552-96848552 on Build GRCh38
rs765211350 Chr.2: 96848477-96848478 on Build GRCh38
rs777007065 Chr.2: 96848754-96848754 on Build GRCh38
rs764278885 Chr.2: 96848394-96848394 on Build GRCh38
rs1003496071 Chr.2: 96848564-96848564 on Build GRCh38
rs994134747 Chr.2: 96848785-96848785 on Build GRCh38
rs565095610 Chr.2: 96848479-96848479 on Build GRCh38
rs921480553 Chr.2: 96848740-96848740 on Build GRCh38
rs1013406367 Chr.2: 96848503-96848504 on Build GRCh38
rs115120038 Chr.2: 96848484-96848484 on Build GRCh38
rs868418103 Chr.2: 96848625-96848625 on Build GRCh38

View Full Product Details