Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFDP1
Entrez Gene ID
10428
Gene Name
craniofacial development protein 1
Gene Aliases
BCNT, BUCENTAUR, CENP-29, CP27, SWC5, Yeti, p97
Chromosome Location
Chr.16: 75293710 - 75433512 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB009285.1 - 6 - 7 917 64
AK226031.1 - 7 - 8 933 64
AK313948.1 - 6 - 7 887 64
BC000991.2 - 6 - 7 941 64
BT009819.1 - 6 - 7 808 64

View Full Product Details