The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006423.2 | NP_006414.2 | 3 - 4 | 451 | 83 |
GenBank mRNA | AF112202.1 | - | 3 - 4 | 381 | 83 |
AF112996.1 | - | 3 - 4 | 377 | 83 | |
AJ133534.1 | - | 3 - 4 | 397 | 83 | |
BC008950.2 | - | 3 - 4 | 403 | 83 | |
BT019964.1 | - | 3 - 4 | 367 | 83 | |
CF242972.1 | - | 3 - 4 | 406 | 83 | |
CR457214.1 | - | 3 - 4 | 367 | 83 | |
CR542116.1 | - | 3 - 4 | 367 | 83 |