The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319676.1 | NP_001306605.1 | 11 - 12 | 1521 | 82 |
NM_006615.2 | NP_006606.1 | 13 - 14 | 1710 | 82 | |
NM_016452.2 | NP_057536.1 | 12 - 13 | 1632 | 82 | |
XM_011544017.1 | XP_011542319.1 | 13 - 14 | 1694 | 82 | |
XM_011544018.1 | XP_011542320.1 | 12 - 13 | 1616 | 82 | |
XM_011544019.2 | XP_011542321.1 | 11 - 12 | 1616 | 82 | |
XM_011544020.1 | XP_011542322.1 | 13 - 14 | 1440 | 82 | |
XM_017000098.1 | XP_016855587.1 | 12 - 13 | 1610 | 82 | |
XM_017000099.1 | XP_016855588.1 | 13 - 14 | 1440 | 82 | |
GenBank mRNA | AB038463.1 | - | 12 - 13 | 1601 | 82 |
AF022799.1 | - | 13 - 14 | 1710 | 82 | |
AK298024.1 | - | 11 - 12 | 1460 | 82 | |
AK309039.1 | - | 14 - 15 | 1624 | 82 | |
AK313629.1 | - | 13 - 14 | 1694 | 82 | |
BC027993.1 | - | 11 - 12 | 1472 | 82 |