Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 131040169-131040657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF4
Entrez Gene ID
50649
Gene Name
Rho guanine nucleotide exchange factor 4
Gene Aliases
ASEF, ASEF1, GEF4, STM6
UniGene
Hs.469935
Gene Chromosome Location
Chr.2: 130836916-131047253 on Build GRCh38
Transcripts
NM_015320.3 NM_032995.2 XM_005263681.1 XM_005263687.3 XM_005263688.2 XM_005263689.4 XM_011511274.2 XM_011511276.2 XM_011511277.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4444741 c.913G>A p.D305N Chr.2: 131040181-131040181 on Build GRCh38
6999319 c.718G>A p.V240I Chr.2: 131040267-131040267 on Build GRCh38
3566458 c.816C>T p.F272F Chr.2: 131040365-131040365 on Build GRCh38
3566456 c.1029C>T p.F343F Chr.2: 131040365-131040365 on Build GRCh38
5491478 c.1059C>T p.R353R Chr.2: 131040395-131040395 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757169701 Chr.2: 131040279-131040279 on Build GRCh38
rs1003649042 Chr.2: 131040238-131040238 on Build GRCh38
rs765515796 Chr.2: 131040244-131040246 on Build GRCh38
rs193061955 Chr.2: 131040528-131040528 on Build GRCh38
rs780673455 Chr.2: 131040371-131040371 on Build GRCh38
rs146115501 Chr.2: 131040338-131040338 on Build GRCh38
rs150910655 Chr.2: 131040398-131040398 on Build GRCh38
rs200228404 Chr.2: 131040472-131040472 on Build GRCh38
rs775393940 Chr.2: 131040401-131040401 on Build GRCh38
rs755900151 Chr.2: 131040323-131040323 on Build GRCh38
rs781194604 Chr.2: 131040453-131040453 on Build GRCh38
rs762165704 Chr.2: 131040225-131040225 on Build GRCh38
rs758494829 Chr.2: 131040253-131040253 on Build GRCh38
rs915397587 Chr.2: 131040352-131040352 on Build GRCh38
rs573997851 Chr.2: 131040508-131040508 on Build GRCh38
rs965919179 Chr.2: 131040629-131040629 on Build GRCh38
rs753539979 Chr.2: 131040422-131040422 on Build GRCh38
rs373443714 Chr.2: 131040217-131040217 on Build GRCh38
rs762428672 Chr.2: 131040305-131040305 on Build GRCh38
rs931711554 Chr.2: 131040400-131040400 on Build GRCh38
rs1049540971 Chr.2: 131040554-131040554 on Build GRCh38
rs776016936 Chr.2: 131040294-131040294 on Build GRCh38
rs1028167718 Chr.2: 131040612-131040612 on Build GRCh38
rs761236894 Chr.2: 131040198-131040199 on Build GRCh38
rs996694376 Chr.2: 131040299-131040299 on Build GRCh38
rs781642969 Chr.2: 131040486-131040486 on Build GRCh38
rs142210980 Chr.2: 131040389-131040389 on Build GRCh38
rs990242485 Chr.2: 131040408-131040408 on Build GRCh38
rs776802297 Chr.2: 131040385-131040385 on Build GRCh38
rs769124274 Chr.2: 131040382-131040382 on Build GRCh38
rs774921903 Chr.2: 131040208-131040208 on Build GRCh38
rs749998864 Chr.2: 131040240-131040240 on Build GRCh38
rs377418807 Chr.2: 131040252-131040252 on Build GRCh38
rs1004886622 Chr.2: 131040582-131040582 on Build GRCh38
rs764930942 Chr.2: 131040232-131040232 on Build GRCh38
rs781105005 Chr.2: 131040303-131040303 on Build GRCh38
rs377371299 Chr.2: 131040374-131040374 on Build GRCh38
rs760551746 Chr.2: 131040404-131040404 on Build GRCh38
rs775933142 Chr.2: 131040213-131040213 on Build GRCh38
rs779084802 Chr.2: 131040348-131040348 on Build GRCh38
rs763885087 Chr.2: 131040410-131040410 on Build GRCh38
rs372799137 Chr.2: 131040202-131040202 on Build GRCh38
rs764357966 Chr.2: 131040333-131040333 on Build GRCh38
rs1038499876 Chr.2: 131040535-131040535 on Build GRCh38
rs752443276 Chr.2: 131040257-131040257 on Build GRCh38
rs759020167 Chr.2: 131040366-131040366 on Build GRCh38
rs773963101 Chr.2: 131040396-131040396 on Build GRCh38
rs755815608 Chr.2: 131040481-131040481 on Build GRCh38
rs143689695 Chr.2: 131040287-131040287 on Build GRCh38
rs765352739 Chr.2: 131040457-131040457 on Build GRCh38
rs759880366 Chr.2: 131040209-131040209 on Build GRCh38
rs757357947 Chr.2: 131040347-131040347 on Build GRCh38
rs754206933 Chr.2: 131040218-131040219 on Build GRCh38
rs996642003 Chr.2: 131040607-131040607 on Build GRCh38
rs752529466 Chr.2: 131040319-131040319 on Build GRCh38
rs956807382 Chr.2: 131040336-131040336 on Build GRCh38
rs370265883 Chr.2: 131040302-131040302 on Build GRCh38
rs375487974 Chr.2: 131040357-131040357 on Build GRCh38
rs750552922 Chr.2: 131040247-131040248 on Build GRCh38
rs777942620 Chr.2: 131040263-131040263 on Build GRCh38
rs1004409437 Chr.2: 131040270-131040270 on Build GRCh38
rs773666383 Chr.2: 131040315-131040315 on Build GRCh38
rs754812631 Chr.2: 131040253-131040253 on Build GRCh38
rs754108609 Chr.2: 131040335-131040335 on Build GRCh38
rs540054630 Chr.2: 131040461-131040461 on Build GRCh38
rs757524332 Chr.2: 131040425-131040425 on Build GRCh38
rs757350348 Chr.2: 131040280-131040280 on Build GRCh38
rs749396177 Chr.2: 131040267-131040267 on Build GRCh38
rs762489457 Chr.2: 131040247-131040247 on Build GRCh38
rs752125154 Chr.2: 131040477-131040477 on Build GRCh38
rs766294491 Chr.2: 131040248-131040248 on Build GRCh38
rs117266463 Chr.2: 131040301-131040301 on Build GRCh38
rs898265920 Chr.2: 131040584-131040584 on Build GRCh38
rs770448187 Chr.2: 131040395-131040395 on Build GRCh38
rs746393820 Chr.2: 131040293-131040293 on Build GRCh38
rs926845320 Chr.2: 131040392-131040392 on Build GRCh38
rs750552483 Chr.2: 131040458-131040458 on Build GRCh38
rs1012668307 Chr.2: 131040271-131040271 on Build GRCh38
rs939554803 Chr.2: 131040511-131040511 on Build GRCh38
rs1056521734 Chr.2: 131040525-131040525 on Build GRCh38
rs749350571 Chr.2: 131040195-131040195 on Build GRCh38
rs554015620 Chr.2: 131040316-131040316 on Build GRCh38
rs764422688 Chr.2: 131040214-131040219 on Build GRCh38
rs779724616 Chr.2: 131040572-131040572 on Build GRCh38
rs376287769 Chr.2: 131040212-131040212 on Build GRCh38
rs780967721 Chr.2: 131040256-131040256 on Build GRCh38
rs767510825 Chr.2: 131040399-131040399 on Build GRCh38
rs1012307051 Chr.2: 131040544-131040544 on Build GRCh38
rs745888592 Chr.2: 131040351-131040351 on Build GRCh38
rs753805026 Chr.2: 131040542-131040542 on Build GRCh38
rs887810102 Chr.2: 131040555-131040555 on Build GRCh38
rs765926582 Chr.2: 131040309-131040309 on Build GRCh38
rs374174739 Chr.2: 131040358-131040358 on Build GRCh38

View Full Product Details