Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 131041139-131041641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF4
Entrez Gene ID
50649
Gene Name
Rho guanine nucleotide exchange factor 4
Gene Aliases
ASEF, ASEF1, GEF4, STM6
UniGene
Hs.469935
Gene Chromosome Location
Chr.2: 130836916-131047253 on Build GRCh38
Transcripts
NM_015320.3 NM_032995.2 XM_005263681.1 XM_005263687.3 XM_005263688.2 XM_005263689.4 XM_011511274.2 XM_011511276.2 XM_011511277.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1006534 c.1111G>A p.D371N Chr.2: 131041236-131041236 on Build GRCh38
309259 c.1141A>T p.N381Y Chr.2: 131041266-131041266 on Build GRCh38
1590528 c.1111G>A p.D371N Chr.2: 131041236-131041236 on Build GRCh38
3566460 c.1233G>A p.M411I Chr.2: 131041358-131041358 on Build GRCh38
3566464 c.1065G>A p.K355K Chr.2: 131041403-131041403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200586233 Chr.2: 131041296-131041296 on Build GRCh38
rs754430232 Chr.2: 131041447-131041447 on Build GRCh38
rs181010119 Chr.2: 131041349-131041349 on Build GRCh38
rs765559711 Chr.2: 131041220-131041220 on Build GRCh38
rs1023900070 Chr.2: 131041342-131041342 on Build GRCh38
rs781521501 Chr.2: 131041253-131041253 on Build GRCh38
rs370743177 Chr.2: 131041180-131041180 on Build GRCh38
rs374604987 Chr.2: 131041430-131041430 on Build GRCh38
rs759687144 Chr.2: 131041359-131041359 on Build GRCh38
rs1052967275 Chr.2: 131041465-131041465 on Build GRCh38
rs147815683 Chr.2: 131041431-131041431 on Build GRCh38
rs375257190 Chr.2: 131041222-131041222 on Build GRCh38
rs746586230 Chr.2: 131041433-131041433 on Build GRCh38
rs770659131 Chr.2: 131041185-131041185 on Build GRCh38
rs542342544 Chr.2: 131041535-131041535 on Build GRCh38
rs185100750 Chr.2: 131041559-131041559 on Build GRCh38
rs111383059 Chr.2: 131041514-131041514 on Build GRCh38
rs1002174451 Chr.2: 131041521-131041521 on Build GRCh38
rs745528155 Chr.2: 131041295-131041295 on Build GRCh38
rs140396073 Chr.2: 131041289-131041289 on Build GRCh38
rs773074596 Chr.2: 131041347-131041347 on Build GRCh38
rs531828961 Chr.2: 131041563-131041563 on Build GRCh38
rs199690025 Chr.2: 131041200-131041200 on Build GRCh38
rs746502583 Chr.2: 131041303-131041303 on Build GRCh38
rs773162642 Chr.2: 131041487-131041487 on Build GRCh38
rs754157280 Chr.2: 131041427-131041427 on Build GRCh38
rs998531092 Chr.2: 131041278-131041278 on Build GRCh38
rs372877378 Chr.2: 131041193-131041193 on Build GRCh38
rs769485434 Chr.2: 131041332-131041332 on Build GRCh38
rs776541125 Chr.2: 131041216-131041216 on Build GRCh38
rs891700955 Chr.2: 131041474-131041474 on Build GRCh38
rs960824712 Chr.2: 131041536-131041536 on Build GRCh38
rs756578048 Chr.2: 131041399-131041399 on Build GRCh38
rs2241460 Chr.2: 131041164-131041164 on Build GRCh38
rs550059915 Chr.2: 131041574-131041574 on Build GRCh38
rs933369712 Chr.2: 131041541-131041541 on Build GRCh38
rs1042881860 Chr.2: 131041613-131041613 on Build GRCh38
rs780691041 Chr.2: 131041464-131041464 on Build GRCh38
rs1011534591 Chr.2: 131041492-131041492 on Build GRCh38
rs777481014 Chr.2: 131041219-131041219 on Build GRCh38
rs539210955 Chr.2: 131041259-131041259 on Build GRCh38
rs745442536 Chr.2: 131041190-131041190 on Build GRCh38
rs753157908 Chr.2: 131041394-131041394 on Build GRCh38
rs748036150 Chr.2: 131041475-131041475 on Build GRCh38
rs760641305 Chr.2: 131041211-131041211 on Build GRCh38
rs889040148 Chr.2: 131041597-131041597 on Build GRCh38
rs367662359 Chr.2: 131041495-131041495 on Build GRCh38
rs992700443 Chr.2: 131041581-131041581 on Build GRCh38
rs908747405 Chr.2: 131041167-131041167 on Build GRCh38
rs368378668 Chr.2: 131041311-131041311 on Build GRCh38
rs768129574 Chr.2: 131041393-131041393 on Build GRCh38
rs372308937 Chr.2: 131041316-131041316 on Build GRCh38
rs778428125 Chr.2: 131041187-131041187 on Build GRCh38
rs767500112 Chr.2: 131041503-131041503 on Build GRCh38
rs372464032 Chr.2: 131041212-131041212 on Build GRCh38
rs751647543 Chr.2: 131041235-131041235 on Build GRCh38
rs775603128 Chr.2: 131041512-131041512 on Build GRCh38
rs923348557 Chr.2: 131041508-131041508 on Build GRCh38
rs138452812 Chr.2: 131041496-131041496 on Build GRCh38
rs751735454 Chr.2: 131041355-131041355 on Build GRCh38
rs779564478 Chr.2: 131041302-131041302 on Build GRCh38
rs149880141 Chr.2: 131041276-131041276 on Build GRCh38
rs115456490 Chr.2: 131041607-131041607 on Build GRCh38
rs766571910 Chr.2: 131041233-131041233 on Build GRCh38
rs544680550 Chr.2: 131041354-131041354 on Build GRCh38
rs756421247 Chr.2: 131041283-131041283 on Build GRCh38
rs752645034 Chr.2: 131041187-131041191 on Build GRCh38
rs560159658 Chr.2: 131041482-131041482 on Build GRCh38
rs909351963 Chr.2: 131041471-131041471 on Build GRCh38
rs756068309 Chr.2: 131041191-131041192 on Build GRCh38
rs556538590 Chr.2: 131041320-131041320 on Build GRCh38
rs759746653 Chr.2: 131041497-131041497 on Build GRCh38
rs375816296 Chr.2: 131041424-131041424 on Build GRCh38
rs776424847 Chr.2: 131041319-131041319 on Build GRCh38
rs763205770 Chr.2: 131041230-131041230 on Build GRCh38
rs749068698 Chr.2: 131041491-131041491 on Build GRCh38
rs879109692 Chr.2: 131041312-131041312 on Build GRCh38

View Full Product Details