Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNNT3
Entrez Gene ID
7140
Gene Name
troponin T3, fast skeletal type
Gene Aliases
TNTF
Chromosome Location
Chr.11: 1919551 - 1938706 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042781.2 NP_001036246.1 14 - 15 891 81
NM_001042782.2 NP_001036247.1 13 - 14 873 81
NM_001297646.1 NP_001284575.1 14 - 15 1131 81
NM_006757.3 NP_006748.1 14 - 15 897 81
XM_006718288.3 XP_006718351.1 9 - 10 924 81
XM_006718290.3 XP_006718353.1 16 - 17 836 81
XM_006718293.2 XP_006718356.1 15 - 16 913 81
XM_006718296.3 XP_006718359.1 14 - 15 800 81
XM_006718299.2 XP_006718362.1 13 - 14 736 81
XM_006718300.3 XP_006718363.1 12 - 13 764 81
XM_006718302.3 XP_006718365.1 11 - 12 746 81
XM_011520343.2 XP_011518645.1 16 - 17 967 81
XM_017018206.1 XP_016873695.1 16 - 17 759 81
XM_017018207.1 XP_016873696.1 15 - 16 747 81
XM_017018208.1 XP_016873697.1 14 - 15 714 81
XM_017018209.1 XP_016873698.1 14 - 15 723 81
XM_017018210.1 XP_016873699.1 13 - 14 688 81
XM_017018211.1 XP_016873700.1 13 - 14 707 81
XM_017018212.1 XP_016873701.1 12 - 13 679 81
XM_017018213.1 XP_016873702.1 11 - 12 664 81
XM_017018215.1 XP_016873704.1 11 - 12 655 81
XM_017018216.1 XP_016873705.1 15 - 16 818 81
XM_017018217.1 XP_016873706.1 14 - 15 797 81
XM_017018218.1 XP_016873707.1 13 - 14 785 81
XM_017018219.1 XP_016873708.1 11 - 12 746 81
GenBank mRNA AJ571879.1 - 1 - 2 48 81
AK056968.1 - 13 - 14 952 81
BC117327.1 - 12 - 13 757 81
BC143537.1 - 12 - 13 757 81
BC171727.1 - 13 - 14 775 81
BC171728.1 - 9 - 10 710 81
BT019997.1 - 13 - 14 679 81
BX640689.1 - 14 - 15 755 81
CR541927.1 - 13 - 14 679 81
DQ778624.1 - 9 - 10 661 81
M21984.1 - 13 - 14 697 81

View Full Product Details