The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006782.3 | NP_006773.2 | 3 - 4 | 377 | 62 |
GenBank mRNA | AF001891.1 | - | 3 - 4 | 359 | 62 |
AF030291.1 | - | 3 - 4 | 364 | 62 | |
AF043611.1 | - | 3 - 4 | 241 | 62 | |
AK291964.1 | - | 3 - 4 | 358 | 62 | |
BC012530.2 | - | 3 - 4 | 312 | 62 | |
BC012814.2 | - | 3 - 4 | 312 | 62 |