Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 143952682-143953181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTDC1
Entrez Gene ID
79712
Gene Name
glycosyltransferase like domain containing 1
Gene Aliases
Hmat-Xa, mat-Xa
UniGene
Hs.44780
Gene Chromosome Location
Chr.2: 143945787-144333092 on Build GRCh38
Transcripts
NM_001006636.3 NM_001164629.3 NM_001284233.1 NM_001284234.1 NM_001284238.1 NM_024659.4 XM_005263774.2 XM_005263782.2 XM_005263784.2 XM_011511843.2 XM_011511855.2 XM_011511856.2 XM_017004917.1 XM_017004918.1 XM_017004919.1 XM_017004920.1 XM_017004921.1 XM_017004922.1 XM_017004923.1 XM_017004924.1 XM_017004925.1 XM_017004926.1 XM_017004927.1 XM_017004928.1 XM_017004929.1 XM_017004930.1 XM_017004931.1 XM_017004932.1 XM_017004933.1 XM_017004934.1 XM_017004935.1 XM_017004936.1 XM_017004937.1 XM_017004938.1 XM_017004939.1 XM_017004940.1 XM_017004941.1 XM_017004942.1 XM_017004943.1 XM_017004944.1 XM_017004945.1 XM_017004946.1 XM_017004947.1 XM_017004948.1 XM_017004949.1 XM_017004950.1 XM_017004951.1 XM_017004952.1 XM_017004953.1 XM_017004954.1 XM_017004955.1 XM_017004956.1 XM_017004957.1 NR_110237.1

Gene Symbol
LOC101928386
Entrez Gene ID
101928386
Gene Name
uncharacterized LOC101928386
Gene Aliases
-
UniGene
Hs.668251
Gene Chromosome Location
Chr.2: 143937067-143964156 on Build GRCh38
Transcripts
NM_001006636.3 NM_001164629.3 NM_001284233.1 NM_001284234.1 NM_001284238.1 NM_024659.4 XM_005263774.2 XM_005263782.2 XM_005263784.2 XM_011511843.2 XM_011511855.2 XM_011511856.2 XM_017004917.1 XM_017004918.1 XM_017004919.1 XM_017004920.1 XM_017004921.1 XM_017004922.1 XM_017004923.1 XM_017004924.1 XM_017004925.1 XM_017004926.1 XM_017004927.1 XM_017004928.1 XM_017004929.1 XM_017004930.1 XM_017004931.1 XM_017004932.1 XM_017004933.1 XM_017004934.1 XM_017004935.1 XM_017004936.1 XM_017004937.1 XM_017004938.1 XM_017004939.1 XM_017004940.1 XM_017004941.1 XM_017004942.1 XM_017004943.1 XM_017004944.1 XM_017004945.1 XM_017004946.1 XM_017004947.1 XM_017004948.1 XM_017004949.1 XM_017004950.1 XM_017004951.1 XM_017004952.1 XM_017004953.1 XM_017004954.1 XM_017004955.1 XM_017004956.1 XM_017004957.1 NR_110237.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4689144 c.1177T>C p.L393L Chr.2: 143952797-143952797 on Build GRCh38
6676658 c.1145A>G p.Y382C Chr.2: 143952829-143952829 on Build GRCh38
3961253 c.1201+2T>C p.? Chr.2: 143952771-143952771 on Build GRCh38
6466286 c.1196T>A p.F399Y Chr.2: 143952778-143952778 on Build GRCh38
275401 c.1177T>G p.L393V Chr.2: 143952797-143952797 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754314829 Chr.2: 143952796-143952796 on Build GRCh38
rs80345971 Chr.2: 143953046-143953046 on Build GRCh38
rs779060431 Chr.2: 143952808-143952808 on Build GRCh38
rs756460666 Chr.2: 143952790-143952790 on Build GRCh38
rs763005367 Chr.2: 143952835-143952835 on Build GRCh38
rs369627508 Chr.2: 143952731-143952731 on Build GRCh38
rs567307761 Chr.2: 143952779-143952779 on Build GRCh38
rs573289184 Chr.2: 143953129-143953129 on Build GRCh38
rs1013387956 Chr.2: 143952989-143952989 on Build GRCh38
rs921606531 Chr.2: 143952891-143952891 on Build GRCh38
rs970194 Chr.2: 143952921-143952921 on Build GRCh38
rs138378289 Chr.2: 143952775-143952775 on Build GRCh38
rs1002523340 Chr.2: 143953057-143953057 on Build GRCh38
rs776485555 Chr.2: 143952732-143952732 on Build GRCh38
rs769680625 Chr.2: 143952830-143952830 on Build GRCh38
rs1032336090 Chr.2: 143953058-143953059 on Build GRCh38
rs994814149 Chr.2: 143952773-143952773 on Build GRCh38
rs762590268 Chr.2: 143952760-143952760 on Build GRCh38
rs770978862 Chr.2: 143952726-143952726 on Build GRCh38
rs751436922 Chr.2: 143952724-143952724 on Build GRCh38
rs150712882 Chr.2: 143952833-143952833 on Build GRCh38
rs769908650 Chr.2: 143952753-143952753 on Build GRCh38
rs977802503 Chr.2: 143952804-143952804 on Build GRCh38
rs890293254 Chr.2: 143953012-143953012 on Build GRCh38
rs774719441 Chr.2: 143952738-143952738 on Build GRCh38
rs931874703 Chr.2: 143952904-143952904 on Build GRCh38
rs144500036 Chr.2: 143953114-143953114 on Build GRCh38
rs1025315129 Chr.2: 143952994-143952994 on Build GRCh38
rs748004099 Chr.2: 143952828-143952828 on Build GRCh38
rs745865976 Chr.2: 143952810-143952810 on Build GRCh38
rs965929752 Chr.2: 143953085-143953085 on Build GRCh38
rs752328833 Chr.2: 143952892-143952892 on Build GRCh38
rs561232875 Chr.2: 143953084-143953084 on Build GRCh38
rs894489600 Chr.2: 143953124-143953124 on Build GRCh38
rs758966120 Chr.2: 143952870-143952870 on Build GRCh38
rs56385884 Chr.2: 143952728-143952728 on Build GRCh38
rs750985783 Chr.2: 143952778-143952778 on Build GRCh38
rs781254115 Chr.2: 143952915-143952915 on Build GRCh38
rs753128174 Chr.2: 143952786-143952786 on Build GRCh38
rs976984801 Chr.2: 143953095-143953095 on Build GRCh38
rs557230571 Chr.2: 143953022-143953022 on Build GRCh38
rs79981290 Chr.2: 143952756-143952756 on Build GRCh38
rs1036088850 Chr.2: 143952930-143952930 on Build GRCh38
rs746121669 Chr.2: 143952729-143952729 on Build GRCh38
rs1036204892 Chr.2: 143953025-143953025 on Build GRCh38
rs190271492 Chr.2: 143952984-143952984 on Build GRCh38
rs764223473 Chr.2: 143953152-143953152 on Build GRCh38
rs767262257 Chr.2: 143952883-143952883 on Build GRCh38
rs773916375 Chr.2: 143952866-143952866 on Build GRCh38
rs148800937 Chr.2: 143952803-143952803 on Build GRCh38
rs142957787 Chr.2: 143952785-143952785 on Build GRCh38
rs988204762 Chr.2: 143953075-143953078 on Build GRCh38
rs761070570 Chr.2: 143952893-143952893 on Build GRCh38
rs552997332 Chr.2: 143952889-143952889 on Build GRCh38
rs180908626 Chr.2: 143953066-143953066 on Build GRCh38
rs770717870 Chr.2: 143952858-143952858 on Build GRCh38
rs201066842 Chr.2: 143953094-143953095 on Build GRCh38
rs778305339 Chr.2: 143952792-143952792 on Build GRCh38
rs377729431 Chr.2: 143952812-143952812 on Build GRCh38
rs780406079 Chr.2: 143952824-143952824 on Build GRCh38
rs140623940 Chr.2: 143952970-143952970 on Build GRCh38
rs370718328 Chr.2: 143952894-143952894 on Build GRCh38
rs989136123 Chr.2: 143952860-143952860 on Build GRCh38
rs760812314 Chr.2: 143953083-143953083 on Build GRCh38
rs890324278 Chr.2: 143952711-143952711 on Build GRCh38

View Full Product Details