Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 143957117-143957622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTDC1
Entrez Gene ID
79712
Gene Name
glycosyltransferase like domain containing 1
Gene Aliases
Hmat-Xa, mat-Xa
UniGene
Hs.44780
Gene Chromosome Location
Chr.2: 143945787-144333092 on Build GRCh38
Transcripts
NM_001006636.3 NM_001164629.3 NM_001284233.1 NM_001284234.1 NM_001284238.1 NM_024659.4 XM_005263774.2 XM_005263782.2 XM_005263784.2 XM_011511843.2 XM_011511855.2 XM_011511856.2 XM_017004917.1 XM_017004918.1 XM_017004919.1 XM_017004920.1 XM_017004921.1 XM_017004922.1 XM_017004923.1 XM_017004924.1 XM_017004925.1 XM_017004926.1 XM_017004927.1 XM_017004928.1 XM_017004929.1 XM_017004930.1 XM_017004931.1 XM_017004932.1 XM_017004933.1 XM_017004934.1 XM_017004935.1 XM_017004936.1 XM_017004937.1 XM_017004938.1 XM_017004939.1 XM_017004940.1 XM_017004941.1 XM_017004942.1 XM_017004943.1 XM_017004944.1 XM_017004945.1 XM_017004946.1 XM_017004947.1 XM_017004948.1 XM_017004949.1 XM_017004950.1 XM_017004951.1 XM_017004952.1 XM_017004953.1 XM_017004954.1 XM_017004955.1 XM_017004956.1 XM_017004957.1 NR_110237.1

Gene Symbol
LOC101928386
Entrez Gene ID
101928386
Gene Name
uncharacterized LOC101928386
Gene Aliases
-
UniGene
Hs.668251
Gene Chromosome Location
Chr.2: 143937067-143964156 on Build GRCh38
Transcripts
NM_001006636.3 NM_001164629.3 NM_001284233.1 NM_001284234.1 NM_001284238.1 NM_024659.4 XM_005263774.2 XM_005263782.2 XM_005263784.2 XM_011511843.2 XM_011511855.2 XM_011511856.2 XM_017004917.1 XM_017004918.1 XM_017004919.1 XM_017004920.1 XM_017004921.1 XM_017004922.1 XM_017004923.1 XM_017004924.1 XM_017004925.1 XM_017004926.1 XM_017004927.1 XM_017004928.1 XM_017004929.1 XM_017004930.1 XM_017004931.1 XM_017004932.1 XM_017004933.1 XM_017004934.1 XM_017004935.1 XM_017004936.1 XM_017004937.1 XM_017004938.1 XM_017004939.1 XM_017004940.1 XM_017004941.1 XM_017004942.1 XM_017004943.1 XM_017004944.1 XM_017004945.1 XM_017004946.1 XM_017004947.1 XM_017004948.1 XM_017004949.1 XM_017004950.1 XM_017004951.1 XM_017004952.1 XM_017004953.1 XM_017004954.1 XM_017004955.1 XM_017004956.1 XM_017004957.1 NR_110237.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1471154 c.1109A>T p.H370L Chr.2: 143957216-143957216 on Build GRCh38
328694 c.1024G>A p.V342I Chr.2: 143957301-143957301 on Build GRCh38
5449514 c.999G>T p.E333D Chr.2: 143957326-143957326 on Build GRCh38
6154295 c.1049G>A p.S350N Chr.2: 143957276-143957276 on Build GRCh38
5545077 c.1022C>T p.S341F Chr.2: 143957303-143957303 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780463193 Chr.2: 143957199-143957199 on Build GRCh38
rs564418470 Chr.2: 143957565-143957565 on Build GRCh38
rs142141358 Chr.2: 143957264-143957264 on Build GRCh38
rs770058000 Chr.2: 143957510-143957510 on Build GRCh38
rs747374187 Chr.2: 143957292-143957292 on Build GRCh38
rs759083628 Chr.2: 143957562-143957562 on Build GRCh38
rs531488769 Chr.2: 143957596-143957596 on Build GRCh38
rs779279246 Chr.2: 143957184-143957184 on Build GRCh38
rs760346860 Chr.2: 143957284-143957284 on Build GRCh38
rs755435166 Chr.2: 143957293-143957293 on Build GRCh38
rs922457056 Chr.2: 143957152-143957152 on Build GRCh38
rs559774351 Chr.2: 143957415-143957415 on Build GRCh38
rs376932571 Chr.2: 143957254-143957254 on Build GRCh38
rs545363612 Chr.2: 143957549-143957549 on Build GRCh38
rs373750631 Chr.2: 143957347-143957347 on Build GRCh38
rs747263965 Chr.2: 143957201-143957201 on Build GRCh38
rs145583666 Chr.2: 143957336-143957336 on Build GRCh38
rs777043369 Chr.2: 143957291-143957291 on Build GRCh38
rs755200476 Chr.2: 143957205-143957205 on Build GRCh38
rs757784103 Chr.2: 143957166-143957166 on Build GRCh38
rs763666412 Chr.2: 143957285-143957285 on Build GRCh38
rs756982881 Chr.2: 143957367-143957367 on Build GRCh38
rs1020419695 Chr.2: 143957382-143957382 on Build GRCh38
rs749166649 Chr.2: 143957365-143957365 on Build GRCh38
rs780767882 Chr.2: 143957216-143957216 on Build GRCh38
rs755257283 Chr.2: 143957340-143957340 on Build GRCh38
rs780805040 Chr.2: 143957392-143957396 on Build GRCh38
rs754140923 Chr.2: 143957153-143957153 on Build GRCh38
rs777725305 Chr.2: 143957225-143957225 on Build GRCh38
rs571708496 Chr.2: 143957489-143957489 on Build GRCh38
rs748271163 Chr.2: 143957476-143957476 on Build GRCh38
rs774945689 Chr.2: 143957275-143957275 on Build GRCh38
rs748970416 Chr.2: 143957239-143957239 on Build GRCh38
rs750862860 Chr.2: 143957321-143957321 on Build GRCh38
rs765819672 Chr.2: 143957301-143957301 on Build GRCh38
rs367821685 Chr.2: 143957551-143957551 on Build GRCh38
rs776240365 Chr.2: 143957546-143957546 on Build GRCh38
rs3731955 Chr.2: 143957194-143957194 on Build GRCh38
rs148297187 Chr.2: 143957338-143957338 on Build GRCh38
rs762068930 Chr.2: 143957294-143957294 on Build GRCh38
rs758895540 Chr.2: 143957334-143957334 on Build GRCh38
rs955011894 Chr.2: 143957251-143957251 on Build GRCh38
rs752501435 Chr.2: 143957349-143957350 on Build GRCh38
rs112900080 Chr.2: 143957560-143957560 on Build GRCh38
rs145121516 Chr.2: 143957247-143957247 on Build GRCh38
rs199617429 Chr.2: 143957147-143957147 on Build GRCh38
rs371230506 Chr.2: 143957196-143957196 on Build GRCh38
rs758921358 Chr.2: 143957192-143957192 on Build GRCh38
rs758384900 Chr.2: 143957197-143957197 on Build GRCh38
rs968498263 Chr.2: 143957469-143957469 on Build GRCh38
rs372716437 Chr.2: 143957244-143957244 on Build GRCh38
rs1005933862 Chr.2: 143957230-143957230 on Build GRCh38
rs778952154 Chr.2: 143957381-143957381 on Build GRCh38
rs910766854 Chr.2: 143957317-143957317 on Build GRCh38

View Full Product Details