The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006919.2 | NP_008850.1 | 7 - 8 | 911 | 70 |
GenBank mRNA | AF428135.1 | - | 6 - 7 | 767 | 70 |
AF428136.1 | - | 6 - 7 | 767 | 70 | |
AF428137.1 | - | 6 - 7 | 767 | 70 | |
AF461146.1 | - | 6 - 7 | 767 | 70 | |
AJ515706.1 | - | 6 - 7 | 797 | 70 | |
AK222746.1 | - | 7 - 8 | 841 | 70 | |
AK222753.1 | - | 7 - 8 | 841 | 70 | |
AK314805.1 | - | 7 - 8 | 841 | 70 | |
AY190327.1 | - | 4 - 5 | 444 | 70 | |
AY245778.1 | - | 6 - 7 | 767 | 70 | |
AY245781.1 | - | 6 - 7 | 767 | 70 | |
BC005224.1 | - | 7 - 8 | 841 | 70 | |
BC008483.1 | - | 7 - 8 | 841 | 70 | |
BT006748.1 | - | 6 - 7 | 767 | 70 | |
EU852041.1 | - | 6 - 7 | 767 | 70 | |
S66896.1 | - | 7 - 8 | 828 | 70 | |
U19556.1 | - | 6 - 7 | 796 | 70 |