The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318877.1 | NP_001305806.1 | 4 - 5 | 358 | 88 |
NM_007003.4 | NP_008934.1 | 4 - 5 | 370 | 88 | |
GenBank mRNA | AF275258.1 | - | 4 - 5 | 356 | 88 |
AI093155.1 | - | 1 - 2 | 88 | ||
AJ005894.1 | - | 4 - 5 | 354 | 88 | |
AL548759.3 | - | 3 - 4 | 321 | 88 | |
BC010897.2 | - | 3 - 4 | 320 | 88 | |
BG435968.1 | - | 4 - 5 | 371 | 88 | |
DA841276.1 | - | 4 - 5 | 352 | 88 |