The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199214.1 | NP_001186143.1 | 1 - 2 | 404 | 64 |
NM_007029.3 | NP_008960.2 | 1 - 2 | 404 | 64 | |
GenBank mRNA | AK292737.1 | - | 1 - 2 | 76 | 64 |
AK297485.1 | - | 1 - 2 | 85 | 64 | |
AK297594.1 | - | 1 - 2 | 98 | 64 | |
AK299500.1 | - | 1 - 2 | 113 | 64 | |
AK308557.1 | - | 1 - 2 | 87 | 64 | |
BC006302.2 | - | 1 - 2 | 73 | 64 | |
BQ069488.1 | - | 1 - 2 | 66 | 64 | |
D50375.1 | - | 1 - 2 | 139 | 64 | |
DC319376.1 | - | 1 - 2 | 404 | 64 | |
S82024.1 | - | 1 - 2 | 50 | 64 |