Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 157736629-157737139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACVR1
Entrez Gene ID
90
Gene Name
activin A receptor type 1
Gene Aliases
ACTRI, ACVR1A, ACVRLK2, ALK2, FOP, SKR1, TSRI
UniGene
Hs.470316
Gene Chromosome Location
Chr.2: 157736446-157875880 on Build GRCh38
Transcripts
NM_001105.4 NM_001111067.2 XM_005246939.3 XM_005246940.3 XM_006712825.3 XM_011512106.2 XM_011512107.2 XM_011512108.2

SNP Detail

SNP ID Chromosome Location
rs898189933 Chr.2: 157736920-157736920 on Build GRCh38
rs763630396 Chr.2: 157736837-157736837 on Build GRCh38
rs764026690 Chr.2: 157736997-157736997 on Build GRCh38
rs886054985 Chr.2: 157737031-157737031 on Build GRCh38
rs550878134 Chr.2: 157736820-157736820 on Build GRCh38
rs569061476 Chr.2: 157736830-157736830 on Build GRCh38
rs1002955476 Chr.2: 157737031-157737031 on Build GRCh38
rs909413916 Chr.2: 157736671-157736671 on Build GRCh38
rs972328613 Chr.2: 157736700-157736700 on Build GRCh38
rs911151956 Chr.2: 157736809-157736809 on Build GRCh38
rs933326394 Chr.2: 157736750-157736750 on Build GRCh38
rs1003501531 Chr.2: 157736713-157736713 on Build GRCh38
rs546726288 Chr.2: 157736726-157736726 on Build GRCh38
rs1012692605 Chr.2: 157737111-157737111 on Build GRCh38
rs892994646 Chr.2: 157736792-157736792 on Build GRCh38
rs761891914 Chr.2: 157736924-157736924 on Build GRCh38
rs893516693 Chr.2: 157737032-157737032 on Build GRCh38
rs74392193 Chr.2: 157736740-157736740 on Build GRCh38
rs188849936 Chr.2: 157736653-157736653 on Build GRCh38
rs946892754 Chr.2: 157737055-157737055 on Build GRCh38
rs983928034 Chr.2: 157736793-157736793 on Build GRCh38
rs1041627116 Chr.2: 157736844-157736844 on Build GRCh38
rs758573643 Chr.2: 157736923-157736923 on Build GRCh38
rs1028134399 Chr.2: 157736779-157736779 on Build GRCh38
rs905418943 Chr.2: 157736689-157736689 on Build GRCh38
rs879007609 Chr.2: 157736982-157736982 on Build GRCh38
rs112908089 Chr.2: 157736769-157736769 on Build GRCh38
rs374226813 Chr.2: 157736951-157736951 on Build GRCh38
rs962342395 Chr.2: 157736743-157736743 on Build GRCh38
rs1051731069 Chr.2: 157736762-157736762 on Build GRCh38
rs549730791 Chr.2: 157736689-157736689 on Build GRCh38
rs761727358 Chr.2: 157737110-157737110 on Build GRCh38
rs570500098 Chr.2: 157736886-157736886 on Build GRCh38
rs1052216154 Chr.2: 157737005-157737005 on Build GRCh38
rs942556250 Chr.2: 157736815-157736815 on Build GRCh38
rs774031962 Chr.2: 157737098-157737098 on Build GRCh38
rs771038233 Chr.2: 157736764-157736764 on Build GRCh38
rs371191871 Chr.2: 157736856-157736856 on Build GRCh38
rs55910418 Chr.2: 157736757-157736757 on Build GRCh38
rs552239569 Chr.2: 157736874-157736874 on Build GRCh38
rs200167578 Chr.2: 157736682-157736682 on Build GRCh38
rs952596523 Chr.2: 157736781-157736781 on Build GRCh38
rs12997 Chr.2: 157736845-157736845 on Build GRCh38

View Full Product Details