Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 157737428-157737925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACVR1
Entrez Gene ID
90
Gene Name
activin A receptor type 1
Gene Aliases
ACTRI, ACVR1A, ACVRLK2, ALK2, FOP, SKR1, TSRI
UniGene
Hs.470316
Gene Chromosome Location
Chr.2: 157736446-157875880 on Build GRCh38
Transcripts
NM_001105.4 NM_001111067.2 XM_005246939.3 XM_005246940.3 XM_006712825.3 XM_011512106.2 XM_011512107.2 XM_011512108.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6956079 c.1450G>T p.A484S Chr.2: 157737611-157737611 on Build GRCh38
1731471 c.1523_1526delACTG p.D508fs? Chr.2: 157737535-157737538 on Build GRCh38
1008039 c.1469G>A p.R490H Chr.2: 157737592-157737592 on Build GRCh38
1008041 c.1467G>A p.L489L Chr.2: 157737594-157737594 on Build GRCh38
6030984 c.1487C>T p.T496I Chr.2: 157737574-157737574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372186308 Chr.2: 157737498-157737498 on Build GRCh38
rs1049312472 Chr.2: 157737796-157737796 on Build GRCh38
rs776697456 Chr.2: 157737499-157737499 on Build GRCh38
rs748669145 Chr.2: 157737491-157737493 on Build GRCh38
rs367857827 Chr.2: 157737552-157737552 on Build GRCh38
rs758395702 Chr.2: 157737492-157737492 on Build GRCh38
rs754926966 Chr.2: 157737664-157737664 on Build GRCh38
rs753946072 Chr.2: 157737654-157737654 on Build GRCh38
rs781143727 Chr.2: 157737673-157737673 on Build GRCh38
rs1026520055 Chr.2: 157737568-157737568 on Build GRCh38
rs777518678 Chr.2: 157737685-157737685 on Build GRCh38
rs145780526 Chr.2: 157737658-157737658 on Build GRCh38
rs770386654 Chr.2: 157737689-157737689 on Build GRCh38
rs113734528 Chr.2: 157737905-157737905 on Build GRCh38
rs1018247049 Chr.2: 157737624-157737624 on Build GRCh38
rs761627707 Chr.2: 157737737-157737737 on Build GRCh38
rs746945657 Chr.2: 157737520-157737520 on Build GRCh38
rs200954410 Chr.2: 157737701-157737701 on Build GRCh38
rs747880953 Chr.2: 157737675-157737675 on Build GRCh38
rs781127382 Chr.2: 157737525-157737525 on Build GRCh38
rs372468278 Chr.2: 157737612-157737612 on Build GRCh38
rs528139252 Chr.2: 157737740-157737740 on Build GRCh38
rs554387941 Chr.2: 157737592-157737592 on Build GRCh38
rs76460538 Chr.2: 157737896-157737896 on Build GRCh38
rs1012967269 Chr.2: 157737514-157737514 on Build GRCh38
rs956790971 Chr.2: 157737623-157737623 on Build GRCh38
rs989403918 Chr.2: 157737655-157737655 on Build GRCh38
rs747858448 Chr.2: 157737536-157737536 on Build GRCh38
rs12936 Chr.2: 157737486-157737486 on Build GRCh38
rs751515337 Chr.2: 157737659-157737659 on Build GRCh38
rs771491043 Chr.2: 157737579-157737579 on Build GRCh38
rs770235094 Chr.2: 157737666-157737667 on Build GRCh38
rs55667327 Chr.2: 157737484-157737484 on Build GRCh38
rs569222994 Chr.2: 157737836-157737836 on Build GRCh38
rs535456170 Chr.2: 157737791-157737791 on Build GRCh38
rs144706255 Chr.2: 157737489-157737489 on Build GRCh38
rs16842009 Chr.2: 157737758-157737758 on Build GRCh38
rs943332918 Chr.2: 157737884-157737884 on Build GRCh38
rs926349740 Chr.2: 157737727-157737727 on Build GRCh38
rs568016148 Chr.2: 157737784-157737784 on Build GRCh38
rs769600810 Chr.2: 157737679-157737679 on Build GRCh38
rs3738927 Chr.2: 157737548-157737548 on Build GRCh38
rs914942419 Chr.2: 157737663-157737663 on Build GRCh38
rs1057002713 Chr.2: 157737760-157737760 on Build GRCh38
rs111369903 Chr.2: 157737479-157737479 on Build GRCh38
rs748946637 Chr.2: 157737686-157737686 on Build GRCh38
rs200076999 Chr.2: 157737642-157737642 on Build GRCh38
rs919522306 Chr.2: 157737761-157737761 on Build GRCh38
rs773899932 Chr.2: 157737695-157737695 on Build GRCh38
rs55788041 Chr.2: 157737555-157737555 on Build GRCh38
rs760430075 Chr.2: 157737832-157737832 on Build GRCh38
rs753003651 Chr.2: 157737608-157737608 on Build GRCh38
rs769751827 Chr.2: 157737554-157737554 on Build GRCh38
rs767830176 Chr.2: 157737606-157737606 on Build GRCh38
rs3738926 Chr.2: 157737803-157737803 on Build GRCh38
rs775226417 Chr.2: 157737790-157737790 on Build GRCh38
rs376866118 Chr.2: 157737522-157737522 on Build GRCh38
rs1057062102 Chr.2: 157737847-157737847 on Build GRCh38
rs757333055 Chr.2: 157737656-157737656 on Build GRCh38

View Full Product Details