The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317962.1 | NP_001304891.1 | 3 - 4 | 342 | 82 |
NM_001317963.1 | NP_001304892.1 | 3 - 4 | 486 | 82 | |
NM_007062.2 | NP_008993.1 | 3 - 4 | 486 | 82 | |
GenBank mRNA | AK223162.1 | - | 3 - 4 | 410 | 82 |
AK290681.1 | - | 3 - 4 | 327 | 82 | |
AK296252.1 | - | 3 - 4 | 343 | 82 | |
BC000067.2 | - | 3 - 4 | 369 | 82 | |
BC001652.2 | - | 3 - 4 | 405 | 82 | |
BC010921.1 | - | 3 - 4 | 420 | 82 | |
BC032127.2 | - | 3 - 4 | 408 | 82 | |
BC040135.1 | - | 3 - 4 | 384 | 82 | |
BC046144.1 | - | 3 - 4 | 400 | 82 | |
DC380488.1 | - | 3 - 4 | 486 | 82 | |
L07758.1 | - | 3 - 4 | 405 | 82 |