The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281771.2 | NP_001268700.1 | 20 - 21 | 3246 | 65 |
NM_001281772.2 | NP_001268701.1 | 23 - 24 | 3826 | 65 | |
NM_001281778.2 | NP_001268707.1 | 21 - 22 | 3627 | 65 | |
NM_001281780.2 | NP_001268709.1 | 22 - 23 | 3695 | 65 | |
NM_001281781.2 | NP_001268710.1 | 20 - 21 | 3348 | 65 | |
NM_001281783.2 | NP_001268712.1 | 21 - 22 | 3702 | 65 | |
NM_001281784.2 | NP_001268713.1 | 20 - 21 | 3486 | 65 | |
NM_012408.5 | NP_036540.3 | 21 - 22 | 3564 | 65 | |
NM_183048.3 | NP_898869.1 | 21 - 22 | 3489 | 65 | |
XM_005260366.2 | XP_005260423.1 | 21 - 22 | 3495 | 65 | |
XM_017027763.1 | XP_016883252.1 | 21 - 22 | 3630 | 65 | |
XM_017027767.1 | XP_016883256.1 | 21 - 22 | 4294 | 65 | |
XM_017027768.1 | XP_016883257.1 | 21 - 22 | 3554 | 65 | |
XM_017027770.1 | XP_016883259.1 | 22 - 23 | 3498 | 65 | |
XM_017027771.1 | XP_016883260.1 | 21 - 22 | 3464 | 65 | |
XM_017027772.1 | XP_016883261.1 | 21 - 22 | 3437 | 65 | |
XM_017027777.1 | XP_016883266.1 | 20 - 21 | 3457 | 65 | |
XM_017027780.1 | XP_016883269.1 | 18 - 19 | 2593 | 65 | |
GenBank mRNA | AB032951.1 | - | 21 - 22 | 3677 | 65 |
AF233453.1 | - | 10 - 11 | 1765 | 65 | |
AF273045.1 | - | 11 - 12 | 2162 | 65 | |
AF454056.1 | - | 21 - 22 | 3381 | 65 | |
AK000275.1 | - | 21 - 22 | 3306 | 65 | |
AK001175.1 | - | 8 - 9 | 1513 | 65 | |
AK294511.1 | - | 23 - 24 | 3636 | 65 | |
BC136608.1 | - | 20 - 21 | 3077 | 65 | |
BC144289.1 | - | 20 - 21 | 3077 | 65 | |
DQ368670.1 | - | 20 - 21 | 3345 | 65 | |
DQ368671.1 | - | 20 - 21 | 3270 | 65 | |
DQ368674.1 | - | 20 - 21 | 3408 | 65 | |
DQ368676.1 | - | 21 - 22 | 3476 | 65 | |
DQ368677.1 | - | 19 - 20 | 3129 | 65 | |
DQ368679.1 | - | 20 - 21 | 3483 | 65 | |
DQ368680.1 | - | 19 - 20 | 3267 | 65 | |
U48251.1 | - | 8 - 9 | 1525 | 65 |