The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012420.2 | NP_036552.1 | 1 - 2 | 236 | 111 |
GenBank mRNA | BC025786.1 | - | 1 - 2 | 215 | 111 |
DC392439.1 | - | 1 - 2 | 236 | 111 | |
U34605.1 | - | 1 - 2 | 122 | 111 |