The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256733.1 | NP_001243662.1 | 7 - 8 | 722 | 93 |
NM_001256735.1 | NP_001243664.1 | 7 - 8 | 692 | 93 | |
NM_001256736.1 | NP_001243665.1 | 7 - 8 | 692 | 93 | |
NM_012446.3 | NP_036578.2 | 8 - 9 | 782 | 93 | |
XM_017009305.1 | XP_016864794.1 | 9 - 10 | 713 | 93 | |
XM_017009306.1 | XP_016864795.1 | 8 - 9 | 697 | 93 | |
XM_017009307.1 | XP_016864796.1 | 7 - 8 | 510 | 93 | |
GenBank mRNA | AF161465.1 | - | 8 - 9 | 612 | 93 |
AK074537.1 | - | 8 - 9 | 577 | 93 | |
AK293544.1 | - | 7 - 8 | 517 | 93 | |
AK294864.1 | - | 7 - 8 | 487 | 93 | |
AK297494.1 | - | 8 - 9 | 490 | 93 | |
AK312337.1 | - | 8 - 9 | 626 | 93 | |
AL080076.1 | - | 8 - 9 | 630 | 93 | |
BC017020.1 | - | 8 - 9 | 630 | 93 | |
BI517313.1 | - | 8 - 9 | 782 | 92 | |
CR936787.1 | - | 7 - 8 | 537 | 93 |